Search
Thermo Scientific Avizo Software functions as a comprehensive digital analytical laboratory for visualizing and understanding imaging data across multiple modalities including microscopy, computed tomography, and spectroscopy. The platform offers specialized editions and extensions that provide advanced capabilities tailored to specific use cases and research requirements. This versatile software enables researchers and engineers to extract meaningful insights from complex imaging data, supporting diverse applications in materials science, industrial inspection, and scientific research.
Thermo Scientific Avizo 3D Pro Software delivers comprehensive numerical characterization of material parameters from imaging data. The platform combines advanced AI image enhancement and segmentation capabilities with traditional analytical approaches to maximize information extraction from complex datasets. This robust software package provides researchers and engineers with an extensive toolset for detailed material analysis, enabling precise quantification of structural and compositional properties across various scientific and industrial applications.
Thermo Scientific Avizo 3D Software for EM Systems is specifically designed for laboratories utilizing electron microscopes, providing specialized capabilities for visualizing and processing electron microscopy imaging and spectroscopy data. The software offers tailored tools that address the unique challenges of EM data analysis, enabling researchers to extract maximum value from their microscopy investments. With dedicated features for handling high-resolution EM datasets, this specialized version of Avizo Software supports advanced visualization, processing, and quantitative analysis of complex microstructural and compositional information across materials science, life sciences, and industrial applications.
Thermo Scientific Avizo 3D Software for Industrial Inspection delivers customizable solutions for routine metrology tasks and challenging measurement applications in industrial settings. Developed through collaborations with major industrial partners, this specialized software can be tailored to meet specific operational requirements across various manufacturing environments. The platform offers adaptable workflows that accommodate the needs of any operator, from basic quality control procedures to complex dimensional analysis. This industrial-focused version of Avizo Software provides manufacturing facilities with powerful inspection capabilities that enhance quality assurance processes while remaining accessible to users at all technical levels.
Thermo Scientific Avizo Trueput Software transforms scanning electron microscopes (SEM) into specialized quality analysis solutions by automating measurements of critical features in microscopy images. The platform features a streamlined user interface and customizable reporting capabilities designed for routine QA/QC tasks in industrial and research environments. This automation enables consistent, operator-independent analysis of microstructural features, enhancing measurement reliability while reducing analysis time. The software's specialized SEM integration provides manufacturing and materials science professionals with an efficient tool for quality control processes requiring high-resolution imaging and precise dimensional measurements.
| Avizo 3D | Avizo 3D Pro | Avizo 3D for Industrial Inspection | Avizo 3D for EM Systems | Subscription program* | |
| Data import, visualization, segmentation, analysis, animation (basic feature set) | |||||
| Advanced segmentation, quantification, EM Toolbox, ToGo Publisher (extended feature set) | Option | ||||
| AI-aided 2D and 3D segmentation | |||||
| Advanced AI features (pretrained models, training in 3D, AI implementation in Segmentation+) | - | ||||
Metrology, reporting, CAD readers (inspection feature set) |
- | - | - | - | |
| FIBStack Wizard, EMWorkflows, Thermo EM Readers, EM Toolbox, Cylinder and Trace Correlation Lines (EM-specific feature set) | - | - | - | - | |
Fiber segmentation and analysis (XFiber extension) |
Option | Option | Option | Option | |
Pore network characterization and modeling (XPNM extension) |
- | Option | Option | Option | |
Advanced meshing and export to solvers (XWind extension) |
Option | Option | Option | Option | |
Material property simulation (XLab Suite extension) |
- | Option | Option | Option | |
Digital volume correlation for material deformation measurements (XDVC extension) |
Option | Option | Option | Option | |
| Localization and visualization of large multichannel and time-series data (Xplore5D extension) | Option | Option | Option | Option | |
Advanced Materials Research for Core Imaging Facilities (XFiber, XWind, XLab , XDVC and XPNM extensions) |
- | Option | Option | Option | - |
Process control environment for PXL FIB-SEM acquisitions on manufacturing (X3D Auto Reconstruction extension) |
- | Option | Option | Option | . |
Co-visualize CT, photography, logs, create virtual logs, reports in digital rock (XCore Profile extension) |
- | Option | - | Option | - |
| Comprehensive digital rock analysis for PNM generation, property simulations, robust meshing (XModelling & Simulation extension) | - | Option | - | Option | - |
*only available for academic and governmental organizations
For Research Use Only. Not for use in diagnostic procedures.