On this page:
High-performance software control platforms to help enhance user experience and data aggregation
Selecting the right equipment is just one step towards creating an efficient workflow. Choosing the right software to complement your equipment and unique workflow needs can make all the difference in your success and time to market. We offer two software platforms to support your process control and data management requirements.
The Thermo Scientific TruBio Bioprocess Control Software—powered on the DeltaV™ Distributed Control Platform from Emerson—facilitates easy integration of multiple vessels to help streamline process control and data management from research to production scale applications—combining robust data transferability, ease of scale-up processes, and intuitive technology.
The Thermo Scientific TruChrom Bioprocess Control Software—available on the DynaChrom single-use chromatography system and powered by DeltaV™—offers easy customization tailored to your processing needs. Designed specifically to control common chromatography and purification skids, this allows for integration and control of downstream processes with transferability of data throughout the workflow.
Watch this video to understand step-by-step single pump calibration with Thermo Scientific TruBio software.
A modern solution that can evolve with your process needs
TruBio Bioprocess Control Software
The TruBio Bioprocess Control Software has features to help drive efficiency and increase stability, including:
  • Integration with the widely trusted automation platform DeltaV™ Live by Emerson
  • Fully scalable control from R&D to production
  • Intuitive design, improved configuration options, and improved data visualization
  • Simplified calibration with easy vessel setup
  • Validated for GMP
  • 21 CFR, Part 11 compatibility
  • No programming or scripting required
User-friendly graphics
  • Graphic is populated directly from the unit info record
  • View 12 tiles on one screen
Process graphic enhancements for ease of use
  • Important modules are easily accessible from the top of the screen
  • Operator can easily enter notes for record keeping
Enhanced user interface for improved data visualization
  • Updated UI/UX interface with intuitive configuration faceplates
  • Dark and light-themed options are available
  • Easy vessel setup
Customization
  • Customized software available
  • Options to easily automate recipes
Thermo Scientific TruChrom Automation Software
The Thermo Scientific TruChrom Automation Software offers built-in sophisticated process control strategies, featuring:
User-friendly configuration
  • Easy user interface that can be configured without having to learn process control programming
  • Quick software switch between different fluid assemblies to adapt operations to new process requirements
Designed to improve accuracy, reduce risk, and enhance process insights
  • Security is built into the recipe parameters
  • Control system with security-level permission to support the safety of your process
  • Recipe safeguards that can stop the process and then continue with the process after the operator checks
A unified upstream and downstream platform
  • Open platform connectivity (OPC) provides system control with integrated solutions into existing plant control systems
  • Allows for simplified batch processing with shared secure files from plant to plant
Research and process development applications
Thermo Scientific TruBio Software with HyPerforma Controllers combine to form one scalable, standard control and automation platform that scales out and up from the clinical stage through production.
Research and process development
Vessel size: 1 L to 30 L vessel size
Controller: HyPerforma G3Lab Controller
Clinical studies
Vessel size: 30 L to 2,000 L vessel size
Controller: HyPerforma G3Lite Controller
Commercial production
Vessel size:
30 L to 5,000 L vessel size
Controller: HyPerforma G3Pro Controller
Single-Use Bioreactors
Our versatile portfolio of S.U.B.s supports robust production across scales and has the flexibility to accommodate a variety of cell lines and processing modalities.
Single-Use Fermentors
A rigorous solution for microbial fermentation applications while featuring the flexibility, ease of use, and efficiency found in single-use systems.
Single-Use Bioprocessing Equipment and Automation
We are committed to delivering exceptional performance, reliability, and quality.
Single-Use Bioprocessing
Our innovative single-use technologies enable you to accelerate biological manufacturing processes to create flexibility and operate more dynamically within upstream and downstream workflows.
BioProcessing Containers and Nalgene Bottles
We offer high-quality storage, transport, and sampling solutions, including Thermo Scientific BioProcess Containers (BPCs) and Thermo Scientific Nalgene containers.
Speak to a specialist
Connect with a Bioprocessing Single-Use Technology representative.
Additional bioprocessing resources
Discover areas where you can cut costs and gain efficiency using our integrated calculator.
Explore our collection of information and insights to support your drug development journey.
Learn, evaluate, connect.
Connect with a Bioprocessing representative or request more info.
For Research Use Only. Not for use in diagnostic procedures.