sho-readyprobes-red-blue
Two-color nuclear staining assay for cell viability
This kit can be used to quickly and easily determine the viability of cells. NucBlue Live reagent stains the nuclei of all cells, while propidium iodide stains only the nuclei of dead cells.
This protocol can be used for:
  • Identifying live and dead cells using a fluorescence microscope
This protocol should not be used for:
  • Flow cytometry
You will need the following for this protocol:
Protocol
1. Culture cells in appropriate medium and vessel for microscopy
2. Add 2 drops each NucBlue Live and propidium iodide per mL of medium to label cells
3. Incubate 5–30 minutes
4. Image cells
Spectral information and storage
 NucBlue LivePropidium iodide
Excitation/Emission360/460 nm535/617 nm
Standard filter setDAPITRITC or RFP
EVOS Light CubeDAPITRITC or RFP
Storage conditionsRoom temperatureRoom temperature
Protocol tips
  • In some cases, more or fewer drops may be needed to achieve optimal staining intensity
  • Image quality may be improved by replacing media with Live Cell Imaging Solution
jurket-R37610
Jurkat cells stained with NucBlue Live reagent and propidium iodide.