Search
T cell processing often involves variable starting materials and hands-on steps that can lead to manufacturing inconsistencies. Dynabeads cell separation technology offers a structured approach to establishing defined isolation and activation, helping teams develop more consistent upstream processes. By supporting controlled early interactions in closed, GMP-ready formats, Gibco CTS Dynabeads magnetic beads help programs build a reliable foundation for later manufacturing stages.
CTS Dynabeads magnetic beads support defined T cell isolation, activation, and depletion by bringing highly specific surface ligands into controlled contact with target cells. Built on a uniform paramagnetic structure, the beads help create steady interaction conditions during early processing steps, supporting reproducible isolation and activation within closed bioprocessing workflows.
| Targeted binding | Active release | Magnetic removal | Buffer exchange and cleanup |
| Single-domain VHH antibodies on the bead surface bind defined CD markers to support selective T cell isolation or activation (1). | After cell separation, the biotin-based Gibco CTS Detachable Dynabeads Release Buffer is added (2) to actively detach beads through a competitive displacement mechanism that releases the biotin derivative on the bead surface (3). | Once detached, CTS Detachable Dynabeads magnetic beads can be magnetically collected and separated from the cell population (4). | Subsequent washes remove the CTS Detachable Dynabeads Release Buffer and remaining components (5), helping generate a highly pure, viable T cell population, including subsets that maintain early memory or stem-like phenotypes. |
Establish reliable early T cell isolation and activation with Dynabeads cell separation technology that supports defined interactions from the start of the process. In addition to isolation and activation, the CTS Dynabeads portfolio includes CD3⁺ cell depletion solutions, expanding support for allogeneic T cell manufacturing workflows that require precise cell population control. The consistent bead composition and well-documented manufacturing framework help reduce cell therapy processing variability between runs and maintain steady operation as workflows evolve. This dependable performance offers programs a stable foundation for moving from development into larger-scale manufacturing.
Some T cell workflows require early activation and controlled bead release for modification and expansion. Gibco CTS Detachable Dynabeads magnetic beads address this need by combining selective binding with the option to release beads at a defined timepoint using the CTS Detachable Dynabeads Release Buffer. This approach helps teams manage activation windows with greater control and move into downstream steps without carrying beads forward. Designed for use in closed, GMP-ready systems, including the CTS DynaCellect Magnetic Separation System, the detachable format supports flexibility across autologous and allogeneic processes.
Established T cell processes often rely on solutions that support predictable activation and steady expansion throughout development. CTS Dynabeads magnetic beads offer a consistent option for programs that use a fixed-activation approach with passive bead release.
Integrate Dynabeads cell separation technology with cell therapy products to support consistent handling from isolation through final processing.
*For Research Use Only. Not for use in diagnostic procedures. Product is a prototype and performance characteristics of this product have not been established.
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.