Search
Our connected solutions include many valuable features that set our offers apart. Explore some of the advantages you can gain when selecting remote monitoring that’s robustly engineered and offers the service and support you need from a life science partner you can trust.
Whether your emphasis is on sample protection, data logging, compliance or asset management, our systems help deliver the peace of mind you want. Designed to accommodate a variety of equipment across locations with varying network requirements, Thermo Scientific Connected Solutions are engineered to be painless to implement and easy to use. Select the system right for you to explore detailed capabilities.
| Remote Monitoring |
Regulatory Compliance |
Asset Performance |
|
|---|---|---|---|
| InstrumentConnect | Smart-Vue Pro | InSight Analytics | Smart-Tracker |
| Availability | |||
| North America | North America and EU | North America | Global |
| Monitoring examples | |||
Temperature Humidity CO2 Power Door open |
Temperature Humidity Differential pressure CO2 |
Temperature Humidity CO2 Ambient temp Energy Consumption Door open Performance |
Temperature Location |
| Equipment Examples | |||
| Ultra-low freezers, freezers, refrigerators, CO2 chambers, baths, cold rooms, cryo systems, incubators, micro incubators, stability chambers, biosafety cabinets | |||
| Compliance | |||
| Reference only | IQ/OQ 21 CFR 11 FDA cGxP |
IQ/OQ 21 CFR 11 |
Reference only |
| Connectivity | |||
Wi-Fi |
LoRa, Bluetooth- cellular/ethernet |
Wi-Fi 915Mhz Bluetooth |
Bluetooth |
| Notification types | |||
| Remote | Local/cloud | Remote | Cloud optional |
| Applications | |||
Mobile Web Cloud |
Mobile Web |
Mobile Web Cloud |
Mobile Cloud |
A single equipment failure or human error can result in the loss of billions of dollars of product or irreplaceable research, so preserving asset integrity is mission critical. That’s why Thermo Fisher Scientific has developed connected solutions to help maintain the integrity of your work whether it’s in transit or in the lab. Did a vaccine in transit maintain required temperature? Was there a power failure risking sample viability? Why waste time worrying, when you can have certainty in knowing the conditions of your assets anytime, anywhere while having freedom to focus on your work and the breakthroughs you seek.
Deploying a quality asset performance system that includes predictive capabilities can serve as a strategic opportunity for capital and operational cost reductions. Analytics that go beyond critical alerts help provide early warning about equipment changes and signal the need for preventative maintenance.
This information can be used to determine useful equipment life and optimize performance, ultimately increasing the return on your equipment investment. Well-maintained equipment can also help reduce the burden on building operations, reducing the demand for energy and HVAC resources, resulting in further operational cost efficiencies. These factors combined can help free up budget for your organization to fund more scientific programs and activities.
“Healthy” freezer operation
Freezer operating with potential for failure
Improve sustainability with InSight analytics
Aging ULT freezers can consume as much energy as a house. Approximately 30-50% may be operating out-of-spec, averaging 10-50% in energy losses. Good data analytics can help identify problems before they put samples at risk, which can help yield cost savings in energy consumption, unplanned maintenance and downtime as well as by helping extend the useful life of the freezer.
Your work is important and complex, requiring intense focus. With Thermo Scientific Connected Solutions, you can eliminate many time-consuming manual data logging and tracking tasks, reduce distracting and potentially disastrous equipment issues, ease demands on staffing resources and focus more energy on your work. Our 21 CFR Part 11 validation compliance system will remotely monitor your equipment, add a layer of sample protection and log data for you in a secure cloud or on premise.
Robust Audit Trail and Documentation
Detailed reporting capabilities, including event, sensor and alarm histories allow for strong documentation and audit trails, eliminating time spent on manual data capturing.
Ensuring the integrity of vaccines, samples and other valuable science doesn’t stop when materials need to be transported to other locations. Thermo Scientific Smart-Tracker is a compact, easy-to-use solution that can help give you confidence that cold chain is maintained close across the globe.
For Research Use Only. Not for use in diagnostic procedures.