Search
The Thermo Scientific TRACE 1600 and Thermo Scientific TRACE 1610 Gas Chromatographs are modular to deliver exceptional versatility for GC and GC-MS applications. Our TRACE 1600 Series gas chromatography systems provide simplified maintenance, reduced downtime, and high flexibility. For optimal flexibility, our suite of iConnect injectors and detectors can be readily shared among multiple TRACE GC systems in numerous configurations to adapt your instruments for priority samples and applications without installation costs. When coupled with our robust injector and detector technology, these gas chromatographs are a smart investment for any size laboratory.
TRACE 1600 GC![]() |
TRACE 1610 GC![]() |
|
|---|---|---|
| Up to two iConnect injectors and two detectors, plus a mass spectrometer on the main frame | ||
| Auxiliary oven for multi-valve and multi-column analyzers | ||
| Methanizer | ||
| Accommodates additional detectors | ||
| iConnect Column Lock connectors | ||
| Helium Saver technology for SSL injector | ||
| Essential local control with single start/stop button and maintenance | ||
| Full instrument control with Chromeleon CDS | ||
| Full local instrument control interface | ||
| 7” HD capacitive touch screen | ||
| Instrument and consumable health tracking with alert notifications for preventive maintenance | ||
| Built-in video maintenance walk-through guides | ||
| Real-time signal monitoring | ||
| Diagnostic functionalities | ||
| Multi-language capabilities | ||
| USB port for touch screen updates and video uploads | ||
| TriPlus 500 Headspace Autosampler controls | ||
| AI/AS 1610 Autosampler controls |