Search
Add objective lenses, Slideport slide inserts, and optical performance monitoring kits to the Thermo Scientific ArrayScan high-content instruments to enhance their functionality and capabilities.
Add a wide range of magnifications using objective lenses for high content instruments.
![]() | Expand the functionality of your Thermo Scientific high-content instrumentAdd a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific ArrayScan HCA Readers and the Thermo Scientific CellInsight NXT High Content Screening (HCS) Platform. |
| Magnification | Objective type | Compatible instrument | Cat. No. |
|---|---|---|---|
| Zeiss 63x (0.75NA) | LD Plan-Neofluar | ArrayScan HCA Readers | N010251 |
| Zeiss 2.5x (0.075NA) | Plan-Neofluar | ArrayScan HCA Readers | N010250 |
| Zeiss 40x (0.75NA) | Plan-Neofluar | ArrayScan HCA Readers | N010099 |
| Zeiss 20x (0.8NA) | Plan-Apochromat | ArrayScan HCA Readers | N010098 |
| Zeiss 40x (0.5NA) | LD A-Plan | ArrayScan HCA Readers | N010097 |
| Olympus 1.25x (0.04NA) | PlanApo N | ArrayScan HCA Readers (not to be used with confocal) | N010125 |
| Olympus 2x (0.08NA) | PlanApo | CellInsight HCS Platforms | NX1002X |
| Olympus 4x (0.16NA) | U Plan S-Apo | CellInsight HCS Platforms | NX1004X |
Monitor and report on changes in the uniformity of fluorescence across an image field on Thermo Scientific High Content Instruments.
The need for monitoring and documenting the consistency of optical system performance relating to field illumination evenness has grown with the demand for production level image-based cell analysis tools. The Thermo Scientific OptiTracker II Optical Performance Monitoring Kit solution is a homogenous fluorescent reagent standard that monitors illumination evenness when used in conjunction with the automated illumination correction wizard on Thermo Scientific High Content Instruments. |
The reporting feature of the illumination correction function on the ArrayScan describes the uniformity of fluorescence across the image field in terms of the average pixel intensity. The OptiTracker Performance Monitoring Kit is a stable cocktail of fluorescent dyes that are active across the entire spectral defection range. Therefore, the performance of the entire optical train from light source to emission filter may be monitored according to a scheduled routine.
| Product | Quantity | Cat. No. | |
|---|---|---|---|
Thermo Scientific OptiTracker II Optical Performance Monitoring Kit
| 1 kit | K1000992 | Request a quote |
Image slides on Thermo Scientific High Content Instruments.
![]() | Image slides on your Thermo Scientific High Content Instrument using Thermo Scientific Slideport Slide Inserts. |
Slideport Slide Inserts have been tested in imaging a wide range of sample types including fixed and live cells, tissue sections, tissue microarrays, and beads. Slideport Slide Inserts allow you to image slides as if you were imaging a standard microplate—any microscope or plate reader that accepts standard microplates will also accept Slideport Slide Inserts. You can easily stack slides and automate imaging, and Slideport Slide Inserts can even be used at a variety of temperatures.
| Product | Quantity | Cat. No. | |
|---|---|---|---|
Thermo Scientific Slideport Slide Inserts
| 1 insert | N0101111 | Request a quote |
Thermo Scientific Slideport Slide Inserts
| 3 inserts | N0101113 | Request a quote |
Thermo Scientific Slideport Slide Inserts
| 5 inserts | N0101115 | Request a quote |