Search
Lithium-ion battery manufacturing presents a set of filtration challenges that should be addressed at each production stage. Particulate contamination in N-Methyl-2-Pyrrolidone (NMP) entering the slurry mixing stage is difficult to correct further downstream. Fine particles that carry through the electrode coating can compromise film uniformity. At the slurry stage, high viscosity, abrasive active materials, and gel particle formation in cathode and anode slurries can create conditions in which filter service life and large-particle-count (LPC) reduction performance can directly affect coating uniformity. At the electrolyte fill stage, chemically aggressive organic carbonate solvent systems and moisture sensitivity may need media with confirmed chemical compatibility and absolute-rated particle control.
Filtration for lithium-ion batteries within the industrial filtration and separation portfolio is organized by process stage, allowing process engineers and quality managers to navigate directly to the stage relevant to their operations.
Lithium-ion battery filtration may require consistent performance across three interrelated capability areas: gel particle and LPC reduction in high-viscosity slurries, precision particle control for electrolyte purity, and operator safety and process containment.
Electrolyte purity is directly linked to cell performance and safety. Particles above the critical size threshold in electrolyte at the fill stage can cause internal short circuits, accelerate capacity fade, and create conditions contributing to thermal runaway in finished cells. Absolute-rated filtration can offer defined particle retention at the micron ratings required for electrolyte applications, with chemical compatibility confirmed for organic carbonate solvent systems.
The Betafine XL filters feature an absolute-rated depth filter constructed of 100% polypropylene with APT, which significantly increases usable filtration surface area while maintaining standard industrial cartridge dimensions.
Electrode slurry is a chemically active, high-viscosity fluid, and filter changeout in slurry service that can introduce operational risk. Open cartridge filter changeout can expose operators to direct contact with active cathode or anode materials and creates contamination risk when the slurry contacts the surrounding environment during service. Changeout-related contamination events affect yield consistency, and operator exposure during filter service is a process design variable with direct operational consequences.
The CTG-Klean system may address both dimensions by containing the cartridge within a sealed pack assembly that is changed out as a complete unit. This enclosed changeout design can minimize operator exposure to slurry materials.
The following products represent the filtration portfolio directly relevant to lithium-ion battery manufacturing.
Product links on this page currently direct to Solventum.com. Product information will soon be available on Thermofisher.com as part of our ongoing integration.
Lithium-ion battery filtration is the application of particulate control across three key manufacturing stages: solvent preparation, electrode slurry coating, and electrolyte filling.
Three primary filtration stages apply in lithium-ion battery manufacturing:
Filtration requirements differ significantly by stage in terms of media selection, viscosity handling, and particle retention targets.
Filter selection for electrode slurry applications involves evaluating slurry viscosity and its effect on differential pressure and flow rate, target particle size for gel particle removal and LPC reduction, filter service life under slurry conditions, chemical compatibility with active cathode or anode materials and carrier solvents, and operator safety during filter changeout. Enclosed cartridge pack systems are designed specifically for high-viscosity slurry applications where open-cartridge changeout creates safety and contamination risks. Connecting with a specialist is recommended for application-specific guidance.