Reliable absorbance results, without complexity
Built on nearly 50 years of trusted Multiskan expertise, the Thermo Scientific Multiskan Ease Absorbance Microplate Reader helps laboratories generate reliable absorbance data quickly, with minimal setup, minimal training, and minimal maintenance.
Designed for routine absorbance assays, Multiskan Ease Absorbance Microplate Reader simplifies everyday workflows so you can focus on results, not instrument complexity. From simple assay setup to immediate data visibility, it supports common applications such as ELISA, cell viability, cytotoxicity, protein quantification and endotoxin testing.
With an intuitive touchscreen interface and preloaded, validated SkanIt Software protocols, lab users can run assays confidently from day one. And with LED-based optics, there’s no lamp replacement, reducing downtime and total cost of ownership. Engineered for straightforward operation and dependable performance, Multiskan Ease Absorbance Microplate Reader reflects Thermo Scientific’s commitment to quality, usability, and global service and support, helping laboratories meet day-to-day research and testing needs with confidence.
Why scientists choose Multiskan Ease Absorbance Microplate Reader
Simple absorbance workflows. Faster, more confident results.
Multiskan Ease Absorbance Microplate Reader reduces common frustrations in routine absorbance testing, complex setup, inconsistent data, long training times, and maintenance downtime. Designed to carry forward the proven and trusted reliability of the legacy Multiskan FC reader and delivers practical improvements that remove everyday barriers in the lab.
You can trust the results
Consistent, reproducible absorbance data
Stable LED optics and hard-coated filters deliver consistent readings across plates, runs, and users.
Reproducibility that holds up over time
Across plates, users, and studies
Controlled incubation and guided workflows maintain reliable results across users and studies.
Fast, repeatable setup
Get started in minutes
Preloaded, validated protocols reduce setup time and minimize user error.
Data is easy to interpret
From plate to answer, faster
Built-in curve fitting and automated reports simplify ELISA and routine data analysis.
High uptime, low maintenance
Ready when your experiments are
LED illumination eliminates lamp replacement, reducing downtime and total cost of ownership.
Easy for every user
Minimal training required
Touchscreen operation and guided software make onboarding fast for any lab member.
Backed by a platform labs trust
Thermo Scientific quality and support
Backed by global service, IQ/OQ options, and proven Multiskan performance.
Fits into modern lab workflows
Flexible and connected
Supports 96- and 384-well plates with incubation, shaking, and USB/Ethernet/cloud connectivity.
Instrument specifications
Technical specifications
Feature
Specification
Overall dimensions (H x W X D)
215 x 295 × 400 mm (8.46 x 11.6 ×15.75 in)
Weight
10 kg (22.05 lb)
Operation conditions
10°C to 40°C; Maximum relative humidity 80% for temperatures up to 31°C. Decreasing linearly to 50% relative humidity at 40°C. Indoor use only
Mains power supply
100–240 Vac, 50/60 Hz, nominal.
Power consumption
Max. 85 W, typical operation (without heating) 25 W
Noise
<50 db (at a distance of 500 mm)
User interface
Standalone use: 8-inch touch screen display
PC control: SkanIt software
Connections
1 USB type A, USB 2.0
1 USB type B, USB 2.0
1 USB type C, USB 2.0
Ethernet port
Plate type
96-well and 384-well microplates, with or without lid, maximum total height 18 mm.
Performance specifications
Absorbance
Light source
White LED
Wavelength range
400–760 nm
Wavelength selection
8-position filter wheel (3 factory-installed, 5 optional)
Factory filters [1]
405 nm, 450 nm and 620 nm
Bandwidth (FWHM)
3–8 nm
Wavelength accuracy
±2 nm
Linearity (Absorbance), at 450 nm
0–3 Abs, 2.0%
Accuracy (Absorbance), at 450 nm
±(1.0% + 0.003 Abs) (0–3 Abs)
Precision (Absorbance), at 450 nm
SD <0.005 Abs or CV <0.5%, whichever is greater
Turbidimetric sensitivity
<5 FTU (formazine turbidimetric units)
Measurement speed [2]
96-well microplates
Fast mode: ≤10 seconds
Precision mode: ≤20 seconds
Incubator
Temperature range From ambient +4°C to 45°C (at room temperature 25°C)
Liquid warm-up time <40 minutes from 25°C to 37°C (96-well microplate, 200 µL water/well)
Mean temperature of wells at 37°C ±0.5°C
Plate temperature uniformity at 37°C <1.0°C
Shaker
Shaking mode
Linear
Shaking speeds
Low (5 Hz, amplitude 15 mm), medium (11 Hz, amplitude 3 mm), high (18 Hz, amplitude 1 mm)
Shaking type
Continuous or pulsed
[1]Hard coated filters with unlimited lifetime.
[2]Minimum kinetic interval time from A1 back to A1.
Ordering information
Catalog # Name Size Price (EUR) Qty
ME001000C Each
7 698,00
Optical filters
Frequently asked questions
Multiskan Ease Absorbance Microplate Reader is used for routine absorbance-based assays, including ELISA, protein quantification, enzyme kinetics, cytotoxicity, and growth curve analysis, delivering accurate and reproducible measurements with minimal training.
Multiskan Ease Absorbance Microplate Reader modernizes the Multiskan FC Photometer with:
  • An LED light source replacing halogen
  • A touchscreen interface replacing a keypad
  • Cloud-ready SkanIt software
  • Built-in incubation and standard 96-/384-well support
It maintains the trusted Multiskan absorbance performance while adding modern usability and connectivity.
Yes. Multiskan Ease Absorbance Microplate Reader is engineered exclusively for absorbance measurements, making it ideal for laboratories that do not require fluorescence or luminescence detection.
Multiskan Ease Absorbance Microplate Reader is ideal for:
  • ELISA-focused laboratories
  • Life science research labs running absorbance assays
  • Teaching and shared labs
  • QC and regulated environments
It’s designed for users who value precision, ease of use, and long-term reliability.
Unlike multimode readers, Multiskan Ease Absorbance Microplate Reader focuses solely on absorbance, eliminating unnecessary complexity while delivering consistent, reproducible results for everyday assays.
Yes. The intuitive touchscreen interface and guided SkanIt workflows allow new users to become productive quickly, even with minimal training.
The Multiskan Ease Absorbance Microplate Reader (Cat. No. ME001000C) includes three factory-installed, hard-coated filters with unlimited lifetime:
  • 405 nm—ALP ELISA with pNPP, ABTS assays, enzyme kinetics, LAL endotoxin assays
  • 450 nm—HRP TMB ELISA, including rapid formats
  • 620 nm—ELISA reference wavelength, Bradford assay alternative
These support many routine applications immediately, without additional purchases.
The Multiskan Ease Absorbance Microplate Reader features an 8-position filter wheel:
  • 3 factory installed filter positions
  • 5 optional positions available for user-selected filters
Wavelength selection: 8-position filter wheel (3 factory-installed, 5 optional).
Yes. The factory installed 405 nm, 450 nm, and 620 nm filters allow immediate use for many of the most common absorbance-based applications, including ELISA and protein quantification assays.
For Research Use Only. Not for use in diagnostic procedures.