Search
Primary XPS region: Be1s
Overlapping regions: Cu3s, Ni3s, Au5s
Binding energies of common chemical states:
| Chemical state | Binding energy Be1s |
|---|---|
| CuxBey, as received | 113.3 eV |
| CuxBey,Ar+ cleaned | 112.5 eV |
Experimental information
Interpretation of XPS spectra
About this element
Symbol: Be
Date of discovery: 1798
Name origin: mineral beryl
Appearance: gray
Discoverer: Nicholas Louis Vauquelin
Obtained from: beryl, chrysoberyl
Melting point: 1560 K
Boiling point: 2742 K
Density[kg/m3]: 1848
Molar volume: 4.85 × 10-6 m3/mol
Protons/Electrons: 4
Neutrons: 5
Shell structure: 2,2
Electron configuration: [He]2s2
Oxidation state: 2
Crystal structure: hexagonal
Beryllium was once known as glucinium, from the Greek word meaning sweet, thanks to the sweet tastes of its salts. It has since been found to be extremely toxic and potentially carcinogenic. Its characteristics as a stiff and lightweight material with stability over a wide temperature range make beryllium ideal for the aerospace industry in the manufacture of lightweight structural materials for high-speed aircraft, missiles, space vehicles, and communication satellites. It is also used in computer equipment, watch springs, and other instruments where light weight and rigidity are needed.
Products
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.