Technical Resources - Media Formulations
21340 - NCTC-109, no Glutamine
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
Glycine75.013.60.18133333
Hydroxy L-proline131.04.090.031221375
L-Alanine89.031.60.3550562
L-Arginine hydrochloride211.031.160.14767772
L-Asparagine-H2O150.09.170.061133333
L-Aspartic acid133.09.910.074511275
L-Cysteine hydrochloride-H2O176.0290.01.6477273
L-Cystine 2HCl313.014.00.044728436
L-Glutamic Acid147.08.260.05619048
L-Histidine hydrochloride-H2O210.026.650.12690476
L-Isoleucine131.018.040.13770993
L-Leucine131.020.440.15603054
L-Lysine hydrochloride183.038.430.21000001
L-Methionine149.04.440.029798659
L-Ornithine HCl169.09.410.055680472
L-Phenylalanine165.016.530.100181825
L-Proline115.06.130.05330435
L-Serine105.010.750.102380954
L-Taurine125.04.180.033439998
L-Threonine119.018.930.15907563
L-Tryptophan204.017.50.085784316
L-Tyrosine disodium salt dihydrate181.021.00.1160221
L-Valine117.025.00.21367522
Vitamins
Biotin244.00.0251.0245902E-4
Choline chloride140.01.250.008928572
D-Calcium pantothenate477.00.0255.2410902E-5
Folic Acid441.00.0255.6689343E-5
Menadione (Vitamin K3)172.00.0251.4534884E-4
Niacinamide122.00.06255.1229505E-4
Nicotinic acid (Niacin)123.00.06255.0813006E-4
Para-Aminobenzoic Acid137.00.1259.1240875E-4
Pyridoxal hydrochloride204.00.06253.0637256E-4
Pyridoxine hydrochloride206.00.06253.0339806E-4
Riboflavin376.00.0256.648936E-5
Thiamine hydrochloride337.00.0257.4183976E-5
Vitamin A (acetate)286.00.258.7412586E-4
Vitamin B121355.010.00.007380074
Vitamin D2 (Calciferol)397.00.256.297229E-4
alpha Tocopherol phos. Na salt554.70.0254.5069406E-5
i-Inositol180.00.1256.9444446E-4
Inorganic Salts
Calcium Chloride (CaCl2) (anhyd.)111.0200.01.8018018
Magnesium Sulfate (MgSO4) (anhyd.)120.097.670.8139166
Potassium Chloride (KCl)75.0400.05.3333335
Sodium Bicarbonate (NaHCO3)84.02200.026.190475
Sodium Chloride (NaCl)58.06800.0117.24138
Sodium Phosphate monobasic (NaH2PO4-H2O)138.0140.01.0144928
Ribonucleosides
5-Methylcytosine225.00.14.4444445E-4
Deoxyribonucleosides
2'Deoxyadenosine251.010.00.03984064
2'Deoxycytidine HCl264.010.00.03787879
2'Deoxyguanosine267.010.00.037453182
Thymidine242.010.00.041322313
Coenzymes
CoA (Coenzyme A)768.02.50.0032552083
DPN (diphosphopyridine nucleotide)663.07.00.01055807
FAD (flavin adenine dinucleotide)786.01.00.0012722646
TPN (triphosphopyridine nucleotide-Na)743.01.00.001345895
TPP Co-carboxylase461.01.00.0021691974
UTP (uridine triphosphate 3Na-2H20)586.01.00.0017064846
Reducing Agents
Ascorbic Acid176.050.00.2840909
Glutathione, monosodium329.010.00.030395137
Other Components
D-Glucosamine HCl216.03.850.017824074
D-Glucose (Dextrose)180.01000.05.5555553
D-Glucuronolactone176.01.80.010227272
L-alpha-Amino-n-butyric acid103.05.50.053398058
Phenol Red376.420.00.053134963
Sodium Acetate136.050.00.36764705
Sodium glucuronate-H2O236.01.80.0076271184
Tween 80®12.5Infinity
Referencia
Related Products: