Technical Resources - Media Formulations
70013 - PBS, 10x, pH 7.2
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Potassium Phosphate monobasic (KH2PO4)136.02100.015.441176
Sodium Chloride (NaCl)58.090000.01551.7241
Sodium Phosphate dibasic (Na2HPO4-7H2O)268.07260.027.089552
Referencia
Related Products:
20012 - PBS, pH 7.2
Cat No: 20012019 , 20012027 , 20012043 , 20012050 , 20012068
10010 - PBS, pH 7.4
Cat No: 10010001 , 10010002 , 10010015 , 10010023 , 10010031 , 10010049 , 10010056 , 10010072
70011 - PBS,10x, pH 7.4
Cat No: 70011036 , 70011044 , 70011051 , 70011069