Technical Resources - Media Formulations
31415 - Leibovitz's L-15
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
Glycine75.0200.02.6666667
L-Alanine89.0225.02.52809
L-Alanyl-L-Glutamine217.0446.02.0552995
L-Arginine174.0500.02.8735633
L-Asparagine132.0250.01.8939394
L-Cysteine121.0120.00.9917355
L-Histidine155.0250.01.6129032
L-Isoleucine131.0250.01.908397
L-Leucine131.0125.00.9541985
L-Lysine146.075.00.51369864
L-Methionine149.075.00.5033557
L-Phenylalanine165.0125.00.75757575
L-Serine105.0200.01.9047619
L-Threonine119.0300.02.5210085
L-Tryptophan204.020.00.09803922
L-Tyrosine181.0300.01.6574585
L-Valine117.0100.00.85470086
Vitamins
Choline chloride140.01.00.007142857
D-Calcium pantothenate477.01.00.002096436
Folic Acid441.01.00.0022675737
Niacinamide122.01.00.008196721
Pyridoxine hydrochloride206.01.00.004854369
Riboflavin 5'-phosphate Na478.00.12.0920503E-4
Thiamine monophosphate442.01.00.0022624435
i-Inositol180.02.00.011111111
Inorganic Salts
Calcium Chloride (CaCl2) (anhyd.)111.0140.01.2612612
Magnesium Chloride (anhydrous)95.093.70.9863158
Magnesium Sulfate (MgSO4) (anhyd.)120.097.670.8139166
Potassium Chloride (KCl)75.0400.05.3333335
Potassium Phosphate monobasic (KH2PO4)136.060.00.44117647
Sodium Chloride (NaCl)58.08000.0137.93103
Sodium Phosphate dibasic (Na2HPO4) anhydrous142.0190.01.3380282
Other Components
D+ Galactose180.0900.05.0
Phenol Red376.410.00.026567481
Sodium Pyruvate110.0550.05.0
Referencia
1. Beall, P.T., Karnaky, K.J., Garretson, L.T. et al. (1983) Biochem. Biophys. Acta 763:19 (rat jejumum).
2. Chinchar, G.D. and Sinclair, J.H. (1978) J. Cell. Physiol. 96:333 (amphibian, frog, ICR, 2A).
3. Flamme, I., Albach, K., Muller, S. et al. (1991) Anat. Record, 229:427 (chick embryos).
4. Foxworthy, P.S. and Eacho, P.I. (1986) Toxicol. Ltrs., 30:189 (Rat hepatocytes).
5. Grothe, C., Otto, D. and Unsicker, K. (1989) Neuroscience 31:649 (Rat embryonic E18 septal neurons).
6. Leibovitz, A. (1963) Am. J. Hygiene 78:173.
7. Matsuda, M. (1983) Develop. Biol., 95:219. (Amphibian [newt] neural plates).
8. Middlebrooks, B.L., Mak, P.C. and Ellender, R.D. (1980) Proc. Soc. Exper. Biol. Med., 165:123 (fish fibroblast).
9. Powell, R.L., Dodson, M.V. and Cloud, J.G. (1989) J. Exper. Zool., 50:333 (Fish Satellite cells, mygenic stem cells).
10. Saad, a.H. and El Ridi, R. (1988) Vet. Immunol Imunopath., 18:369 (reptile [lizard] spleen cells).
11. Tate, J.M., Oberpriller, J.O. and Oberpriller, J.C. (1989) Tissue and Cell, 21:335 (Newt cardiac monocytes).
Related Products:
11415 - Leibovitz's L-15
Cat No: 11415049 , 11415056 , 11415064 , 11415114