Search
Neurobasal™ Media are basal media formulated to meet the neuronal cells special requirements.
They allow for long-term maintenance of the normal phenotype and growth of neuronal cells, and maintain pure populations of neuronal cells without the need of an astrocyte feeder layer. The concentration of NaCl is adjusted to bring the Osmolality to 260 +/- 10 mOsm
Catalog Number(s)
10888022| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| Glycine | 75.0 | 30.0 | 0.4 |
| L-Alanine | 89.0 | 2.0 | 0.02247191 |
| L-Arginine hydrochloride | 211.0 | 84.0 | 0.39810428 |
| L-Asparagine-H2O | 150.0 | 0.83 | 0.0055333334 |
| L-Cysteine | 121.0 | 31.5 | 0.2603306 |
| L-Histidine hydrochloride-H2O | 210.0 | 42.0 | 0.2 |
| L-Isoleucine | 131.0 | 105.0 | 0.8015267 |
| L-Leucine | 131.0 | 105.0 | 0.8015267 |
| L-Lysine hydrochloride | 183.0 | 146.0 | 0.7978142 |
| L-Methionine | 149.0 | 30.0 | 0.20134228 |
| L-Phenylalanine | 165.0 | 66.0 | 0.4 |
| L-Proline | 115.0 | 7.76 | 0.06747826 |
| L-Serine | 105.0 | 42.0 | 0.4 |
| L-Threonine | 119.0 | 95.0 | 0.79831934 |
| L-Tryptophan | 204.0 | 16.0 | 0.078431375 |
| L-Tyrosine | 181.0 | 72.0 | 0.39779004 |
| L-Valine | 117.0 | 94.0 | 0.8034188 |
| Vitamins | |||
| Choline chloride | 140.0 | 4.0 | 0.028571429 |
| D-Calcium pantothenate | 477.0 | 4.0 | 0.008385744 |
| Folic Acid | 441.0 | 4.0 | 0.009070295 |
| Niacinamide | 122.0 | 4.0 | 0.032786883 |
| Pyridoxal hydrochloride | 204.0 | 4.0 | 0.019607844 |
| Riboflavin | 376.0 | 0.4 | 0.0010638298 |
| Thiamine hydrochloride | 337.0 | 4.0 | 0.011869436 |
| Vitamin B12 | 1355.0 | 0.0068 | 5.0184503E-6 |
| i-Inositol | 180.0 | 7.2 | 0.04 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 200.0 | 1.8018018 |
| Ferric Nitrate (Fe(NO3)3"9H2O) | 404.0 | 0.1 | 2.4752476E-4 |
| Magnesium Chloride (anhydrous) | 95.0 | 77.3 | 0.8136842 |
| Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 2200.0 | 26.190475 |
| Sodium Chloride (NaCl) | 58.0 | 4000.0 | 68.965515 |
| Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 125.0 | 0.9057971 |
| Zinc sulfate (ZnSO4-7H2O) | 288.0 | 0.194 | 6.736111E-4 |
| Other Components | |||
| D-Glucose (Dextrose) | 180.0 | 4500.0 | 25.0 |
| HEPES | 238.0 | 2600.0 | 10.92437 |
| Phenol Red | 376.4 | 8.1 | 0.021519661 |
| Sodium Pyruvate | 110.0 | 25.0 | 0.22727273 |
NEUROBASAL™ Medium (1X) liquid
Cat No:
21103049
Neurobasal™-A Medium (1X) Liquid without Phenol Red
Cat No:
12349015
Neurobasal™ Medium (1X) Liquid without Phenol Red
Cat No:
12348017