On this page:
Automate and streamline magnetic separation
Manual magnetic separation steps can be time-consuming and difficult to standardize across operators and scales. The Gibco CTS DynaCellect Magnetic Separation System is designed to automate these tasks within a closed, benchtop system that supports consistent operation, helps reduce manual touchpoints, and facilitates reproducibility across T cell therapy workflows.
Instrument
CTS DynaCellect Magnetic Separation System
  • Offers high cell purity, recovery, and viability 
  • Closed system easily integrates into existing processes
  • Gibco Cell Therapy Systems (CTS) product manufactured under GMP guidelines
Software
CTS DynaCellect Magnetic Separation System Software
  • Intuitive and easy-to-use embedded graphical interface (eGUI) 
  • Control magnet engagement, pinch valves, pump speed, and rocker angles
  • Available software upgrade that supports compliance with 21 CFR Part 11
Features of CTS DynaCellect Magnetic Separation System
The CTS DynaCellect Magnetic Separation System supports efficient magnetic bead-based cell handling with processing speed, flexibility, and scalability suited for T cell therapy manufacturing. Its connected design—used with the Gibco CTS DynaCellect Cell Isolation Kit, Gibco CTS DynaCellect Bead Removal Kit, and programmable software—helps standardize steps while supporting reproducible performance across cell therapy processes.
Enhance processing speed
CTS DynaCellect Magnetic Separation System supports isolation of up to 1 L of starting material in approximately 70–100 minutes, depending on pump settings, and enables continuous-flow bead removal for controlled, rapid processing. When used with Gibco CTS Dynabeads CD3/CD28 beads, bead removal can be performed at 50 mL/min with high recovery, with a pump capacity up to 150 mL/min for process development needs.
Scale cell isolation and bead removal
Scale T cell manufacturing workflows with the CTS DynaCellect Magnetic Separation System, which supports 50 mL to 1 L isolation volumes for both autologous and allogeneic processes. Its continuous-flow bead removal method allows for larger working volumes (up to 10 L), helping sustain cell viability and recovery while minimizing residual beads during scale transitions. Scalable cell isolation and magnetic bead removal are enabled through sterile, single-use kits manufactured under GMP guidelines.
Adapt protocols for diverse workflows
Magnetic separation workflows can adapt with programmable control over isolation and bead-removal steps. The system includes two ready-to-run protocols and the option to create custom protocols to support different cell types and process needs. Users can adjust magnet engagement, pump speed, pinch valves, and rocker settings, supporting flexibility from early development through clinical manufacturing.
Explore the CTS DynaCellect Magnetic Separation System 3D interactive tour
Interact with a detailed view of the system to see how its components, user interface, and process flow work together in a closed, automated format.
Empower control with the clinical manufacturing mode
Strengthen process control during scale-up by using the clinical manufacturing mode available for the CTS DynaCellect Magnetic Separation System. This mode locks optimized protocols to support consistent operation, and an optional software module supports 21 CFR Part 11-aligned record-keeping with configurable user roles, audit logging, and e-Signature workflows managed through the SAE Administrator Console. A licensed installation is completed by a field service engineer (FSE).
Evaluate performance in T cell isolation and activation
The CTS DynaCellect Magnetic Separation System helps users easily scale from research to clinical and commercial manufacturing. This highly scalable system uses a continuous flow process for bead removal, allowing for easy scaling, verified from 50 mL to 10 L of reaction volume (with optimal total target cell range of 108 to 1010) for cell isolation.
Assess enrichment outcomes using the CTS DynaCellect Magnetic Separation System with CTS Dynabeads CD3/CD28 beads.
Figure 1. Cell phenotype of the starting material and enriched target cells after isolation. (A) The proportion of target cells in the unwashed starting material was ~53.6%. (B) After isolation using CTS Detachable Dynabeads CD3/CD28 and the CTS DynaCellect Magnetic Separation System, the purity of the target T cells was ~97.7% as measured by flow cytometry.
Review isolation efficiency and viability performance supported by the magnetic system.
Figure 2. High isolation efficiency with no impact on cell viability. Using CTS Detachable Dynabeads CD3/CD28 and the CTS DynaCellect Magnetic Separation System, (A) the average isolation efficiency was 92.8% (n = 4), and (B) the viability of isolated cells was slightly increased across bead release days 1 through 3, as determined by flow cytometry.
Track activation marker expression following isolation with the system.
Figure 3. Confirmed activation of isolated target cells. CTS Detachable Dynabeads CD3/CD28 were released on days 1, 2, and 3 after isolation. The early activation marker CD69 was expressed on target cells on day 1 and was downregulated by day 3. The late activation marker CD25 was highly expressed on day 1 and continued to be upregulated through day 3. The expression profile adheres to expected target cell kinetics (n = 4).
System specifications
Support consistent operation across development and manufacturing with the CTS DynaCellect Magnetic Separation System, produced in a GMP and ISO-certified facility. The system can function as a standalone instrument or integrate into closed cell therapy workflows, with scalable operation that spans small development volumes through larger clinical and commercial needs.
Table 2. Instrument specifications
Processing time
~70–100 minutes
Efficiency
>88% isolation (with CTS Detachable Dynabeads CD3/CD28)
DeltaV compatibility (OPC-UA)
Yes
Reproducibility
Coefficient of variation (CV) of 3%
Purity
97% (with CTS Detachable Dynabeads CD3/CD28)
Viability
Unaffected
Isolation scale
Maximum 1,000 mL working volume (equivalent to ~1010 T cells as input)
Bead removal scale
Verified up to 10 L
Maximum cell input
~1010 T cells (1,000 mL isolation volume)
Pump speed
1–150 mL/min
Rocker angle
-30° to 30°
Rocker speed
1–30 cpm (cycles per minute)
Maximum air pressure
2 bar
Supports compliance with 21 CFR Part 11
Yes, with upgrade
Open Platform Communications–Unified Architecture (OPC-UA) compatible
Yes
Cloud connect utility
Yes
Touch display
20.32 cm
Voltage
100–240 V AC ±10%, 50/60 Hz, 600 W
Weight
50 kg
Depth
562 mm
Width
749.5 mm
Height
485 mm with pole
Advance T cell therapies with accessories and consumables
Extend the capabilities of the CTS DynaCellect Magnetic Separation System with single-use consumables for isolation, depletion, and bead removal in your workflows.
CTS DynaCellect Cell Isolation Kit
Support closed magnetic separation workflows with the CTS DynaCellect Cell Isolation Kit, designed for immune cell isolation and activation with minimal manual handling. The kit includes weldable tubing and sterile single-use components to help maintain reproducibility across process runs.
CTS DynaCellect Bead Removal Kit
Enable controlled and efficient bead detachment with the CTS DynaCellect Bead Removal Kit. Its single-use format supports rapid bead removal following activation while helping preserve cell viability across different scales.
CTS OpTmizer T cell culture media
Use CTS OpTmizer media and related formulations to support consistent T cell expansion and viability across automated workflows. These media solutions are produced in GMP and ISO-certified facilities and are suitable for integration with closed cell therapy bioprocessing systems.
CTS Detachable Dynabeads Release Buffer
Incorporate compatible reagents, such as CTS Detachable Dynabeads Release Buffer, to support bead removal and downstream processing using CTS DynaCellect Magnetic Separation System. These reagents are designed for use in bioprocessing environments that require closed, controlled operation.
Designed for flexible integration across T cell workflows
The CTS DynaCellect Magnetic Separation System is purpose-built for T cell therapy manufacturing. The system integrates with Gibco CTS Dynabeads magnetic beads to enable efficient T cell isolation, activation, and bead removal. The design fits both autologous and allogeneic workflows, supporting smooth transitions from early process development to clinical-scale production.
Enhance cell therapy workflows with modular solutions
Integrate the CTS DynaCellect Magnetic Separation System with other cell therapy products to support T cell processing.
CTS DynaMag Magnet
Support small-scale or development-stage magnetic separation with the CTS DynaMag Magnet, designed for manual bead-based workflows that complement automated processing.
CTS Rotea Counterflow Centrifugation System
Help automate washing, concentration, and separation steps with the CTS Rotea Counterflow Centrifugation System, which integrates easily into closed-cell therapy manufacturing processes.
Software
Configure magnetic separation steps using the system’s user-programmable software, which allows you to create custom protocols with the Protocol Builder. The instrument also offers two standard protocols, one for cell isolation and one for bead removal. Users can adjust magnet engagement, pump speed, rocker settings, and valve positions to meet process needs. Once optimized, protocols can be locked in clinical manufacturing mode, with optional features that support 21 CFR Part 11-aligned record-keeping, audit logging, user-level permissions, and summary file generation.
System services
Maintain reliable CTS DynaCellect Magnetic Separation System operation with global service coverage, warranty options, and qualification support to help keep your instrument performing as expected throughout its lifecycle. More than 2,000 trained professionals make up our service and support network, ready to assist you when you need it. Each system comes with a standard 1-year warranty, which covers costs for travel or shipping, labor, and parts for repairs.
AB Assurance Plan
Planned maintenance
Receive a planned maintenance service visit from the FSE as part of the AB Assurance Plan during the second year. During this visit, the engineer inspects the system, performs software updates as needed, cleans and lubricates components, completes calibration and functional testing, and documents findings to help you prevent downtime, reduce strain on your staff, and help extend the life of your instrument.
Purchasing service plans
Select a service plan at the time of your purchase or add an extended warranty later, depending on regional availability. Customers may contact their sales representative for guidance on options that allow additional coverage or extend support for their CTS DynaCellect Magnetic Separation System.
Qualification services
Add instrument hardware qualifications, such as Installation Qualification (IQ) and Operational Qualification (OQ), to document system installation and verify that subsystems operate within manufacturer specifications. These services are recommended during installation, after major maintenance, following an instrument move, or after software or hardware updates, with audit-ready documentation by qualification specialists.
Frequently asked questions
The CTS DynaCellect Magnetic Separation System is designed to automate bead-based magnetic separation steps for T cell isolation and activation. It supports cell isolation and bead removal in a closed, automated format, helping maintain process consistency and operator efficiency. The system is designed for use with both CTS Dynabeads magnetic beads and CTS Detachable Dynabeads magnetic beads to support controlled magnetic separation across manufacturing stages.
The CTS DynaCellect Magnetic Separation System supports both autologous and allogeneic T cell therapy workflows through its scalable isolation volumes and continuous-flow bead process. Users can adapt protocols to different starting materials and batch sizes, enabling instrument use for both small autologous runs to larger manufacturing formats used in allogeneic processes.
The CTS DynaCellect Magnetic Separation System is designed for use in closed, automated workflows that support GMP-aligned bioprocessing. Its single-use consumables, weldable tubing, and programmable software help maintain process control, reduce manual handling, and support electronic record-keeping when used with the optional 21 CFR Part 11 software module.
Discover cell therapy bioprocessing solutions
Explore cell therapy bioprocessing solutions that help maintain consistency as programs progress from development to manufacturing and adapt to autologous or allogeneic needs.
Cell therapy resources
Explore cell therapy resources for in-depth workflow guidance, application notes, and technical materials that support T cell processing from early development through manufacturing.
Enhance cell therapy manufacturing with the CTS DynaCellect Magnetic Separation System
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.