Search
At harvest, upstream decisions become downstream constraints. Rising cell densities and intensified titers increase loads of cell debris and dissolved impurities, including host cell protein (HCP) and DNA, that must be managed before purification begins. Clarification choices at this stage directly affect feedstream quality, column performance, and scalability.
At Thermo Fisher Scientific, we work with process development teams to evaluate harvest within the broader context of downstream performance and scale.
Clarification at harvest is addressed using strategies defined by their separation mechanism and role within the workflow. Depth filtration relies mostly on size-based retention and mechanical entrapment to remove cells and debris and is commonly used to manage high solids loads. Chromatographic clarification uses charge-functionalized media to retain negatively charged soluble and insoluble impurities while the target molecule flows through. Applied to direct harvest or centrate, these approaches shape how clarification is implemented across traditional, next-generation intensified, and polishing clarification workflows.
Evaluate harvest and clarification solutions by comparing how different approaches address solids load, impurity profiles, workflow integration, and scalability.
We assess how these factors influence consistent performance from development through manufacturing.
Table 1. Select the right harvest and clarification solution
| Traditional clarification | Enhanced clarification | Intensified clarification | |
|---|---|---|---|
| Solution | Zeta Plus Depth Filters | Emphaze AEX Hybrid Purifier | Harvest RC Chromatographic Clarifier platform |
| Technology platform | Cellulose-based depth filtration media | Q-functionalized polypropylene nonwoven with integrated 0.2 µm membrane | Q-functionalized polypropylene nonwoven (fiber-based AEX) with 0.2 µm membrane |
| Primary mechanism | Size-based retention with charge-assisted entrapment | AEX chromatography with size exclusion | AEX chromatography (charge-based separation) |
| Materials | Cellulose, binder resin, filter aid | Synthetic | Synthetic |
| Position in workflow | Primary clarification of direct harvest or centrate | Used after depth filtration; may replace second-stage depth filtration | Direct harvest or centrate clarification in an intensified approach |
| When to use | When managing high solid loads using established clarification strategies | When improving effluent quality from depth filtration and protecting downstream capture | When simplifying clarification trains and intensifying processes |
| Scalability | Scalability from lab to multi-capsule production scale | Linear scalability from lab to multi-capsule commercial production | Linear scalability from lab to multi-capsule commercial production |
| Water-flushable | Yes | No | Yes |
Clarified feedstream quality influences downstream chromatography performance, including column capacity and pressure behavior, while helping protect membranes and sterile filters from fouling. Well-designed clarification steps help stabilize downstream performance and reduce batch-to-batch variability.
Centrifugation may be evaluated in processes with very high cell densities or large volumetric throughputs where bulk solids removal is needed upfront. It is often used to reduce the particulate burden before downstream clarification, helping limit depth filter sizing or capacity requirements. Centrifugation is typically considered as part of a hybrid workflow.
Depth filtration relies primarily on size-based capture and mechanical entrapment to remove cells and debris throughout a tortuous porous media matrix. As solids accumulate, flow resistance and pressure differentials may increase, influencing process performance in shear-sensitive systems.
Fiber-based chromatographic clarification uses charge-based interactions to retain cells and soluble impurities while allowing the target molecule to flow through. Each approach is selected based on solids load, impurity profile, and integration with downstream purification steps.
Scalability is influenced by feedstream variability, solids load, and throughput requirements as processes advance from development to manufacturing. Footprint constraints, single-use formats, and predictable scale-up behavior are also key considerations. Designing clarification steps that support clarified fluid quality across volumes helps enable reliable technology transfer.
High cell-density processes increase solids, DNA, and soluble impurity loads, requiring clarification strategies that handle higher particulate burdens without compromising effluent quality. Optimization focuses on using clarification approaches that maintain throughput, reduce shear, and protect downstream operations. This may include staged clarification, charge-based separation mechanisms, or workflow designs that reduce reliance on multiple unit operations at scale.
Intended use of the products mentioned on this page vary. For specific intended use statements, please refer to the Instructions for Use for the product.