Search
It is sometimes possible to stain cells after fixation but this is dependent upon the effects of fixation on the epitope of your protein-of-interest. The list below contains information about the performance of different clones after fixation using our Invitrogen™ eBioscience™ Intracellular Fixation & Permeabilization Buffer Set (Cat. No. 88-8824) or after IC Fixation Buffer (Cat. No. 00-8222) followed by treatment with methanol. Please note that staining after fixation may allow for intracellular stores of protein to also be stained with the antibody.
| Antigen | Clone | Live cells (before IC fixation)1 | After IC fixation and perm wash1 | After IC fixation/ methanol2 |
|---|---|---|---|---|
| HUMAN | ||||
| CD3 | OKT3 | +++ | ++ | + |
| UCHT1 | +++ | +++ | +++ | |
| SK7 | +++ | ++ | +++ | |
| CD4 | OKT4 | +++ | + | +/- |
| RPA-T4 | +++ | +++ | + | |
| SK3 | +++ | +++ | + | |
| CD5 | UCHT2 | +++ | +++ | +++ |
| CD8 | OKT8 | +++ | +/- | - |
| RPA-T8 | +++ | +++ | +/- | |
| SK1 | +++ | +++ | +/- | |
| CD8b | SIDI8BEE | +++ | ++ | ++ |
| CD11a | HI111 | +++ | +++ | + |
| CD11b | CBRM1/5 | + | - | +++ |
| ICRF44 | ++ | - | ++ | |
| CD11c | 3.9 | ++ | + | ++ |
| CD14 | 61D3 | +++ | +++ | ++ |
| CD19 | HIB19 | +++ | ++ | +/- |
| CD20 | 2H7 | +++ | +++ | - |
| CD25 | BC96 | + | - | +/- |
| CD27 | O323 | +++ | + | + |
| CD31 | WM59 | +++ | ++ | +++ |
| CD33 | p67.6 | +++ | +++ | +++ |
| CD38 | HB7 | ++ | - | ++ |
| HIT2 | ++ | - | ++ | |
| CD40 | 5C3 | ++ | - | ++ |
| CD44 | IM7 | +++ | +++ | +++ |
| CD45 | 2D1 | +++ | +++ | +++ |
| HI30 | +++ | +++ | +++ | |
| CD45RA | HI100 | +++ | +++ | +++ |
| CD45RO | UCHL1 | +++ | +++ | ++ |
| CD56 | MEM-188 | + | - | +/- |
| CB56 | + | - | +/- | |
| CMSSB | ++ | +/- | - | |
| CD57 | TB01 | ++ | ++ | ++ |
| CD62L | DREG-56 | ++ | - | ++ |
| CD69 | FN50 | +++ | ++ | ++ |
| CD80 | 2D10.4 | + | - | + |
| CD83 | HB15e | ++ | ++ | ++ |
| CD86 | IT2.2 | + | + | + |
| CD94 | HP-3D9 | ++ | ++ | ++ |
| CD95 | DX2 | + | - | + |
| CD127 | RDR5 | ++ | + | ++ |
| CD161 | HP-3G10 | ++ | ++ | + |
| MOUSE | ||||
| CD3 | 145-2C11 | ++ | + | +/- |
| 500A2 | +++ | +++ | +++ | |
| 17A2 | +++ | +++ | +++ | |
| CD4 | GK1.5 | +++ | ++ | +++ |
| RM4-5 | +++ | +++ | +++ | |
| CD8 | 53-6.7 | +++ | ++ | ++ |
| CD11b | M1/70 | ++ | ++ | ++ |
| CD11c | N418 | +++ | +++ | ++ |
| CD19 | 1D3 | +++ | ++ | - |
| MB19-1 | ++ | - | - | |
| CD24 | M1/69 | +++ | +++ | ++ |
| CD25 | PC61.5 | + | - | + |
| 3C7 | ++ | - | + | |
| 7D4 | ++ | ++ | + | |
| CD39 | 24DMS1 | + | - | ++ |
| CD44 | IM7 | ++ | ++ | ++ |
| CD45 | 30-F11 | +++ | +++ | +++ |
| CD45R(B220) | RA3-6B2 | +++ | ++ | +++ |
| CD49b | DX5 | ++ | - | + |
| CD69 | H1.2F3 | ++ | - | - |
| CD185(CXCR5) | SPRCL5 | ++ | - | - |
| GR-1 | RB6-8C5 | +++ | +++ | ++ |
| NK1.1 | PK136 | ++ | + | + |
| IgD | 11-26c | +++ | +++* | ++ |
| IgM | ll/41 | ++ | ++ | ++ |
| TCR beta | H57-597 | ++ | ++ | ++ |
| RAT | ||||
| CD25 | OX39 | + | - | |
| CANINE | ||||
| CD4 | ykix302.9 | +++ | + | |
| CD5 | ykix322.3 | +++ | - | |
| CD8a | ycate55.9 | +++ | ++ | |
| CD44 | ykix337.8 | +++ | ++ | |
| CD45 | ykix716.13 | +++ | ++ | |
| CD45R | ykix753.22 | +++ | ++ | |
| CD90 | ykix337.217 | +++ | ++ | |
| MHC Class II | ykix334.2 | +++ | +++ |
1.Testing carried out with eBioscience Fixation & Permeabilization Kit for 15–30 minutes at room temperature, followed by a perm wash. See Staining Intracellular Antigens for Flow Cytometry, Protocol A.
*Please note: Clones marked with an asterisk (*) indicate that longer fixation times are detrimental to staining.
2.Testing carried out for 15–30 minutes at room temperature in 50% IC fixation, followed by a 30-minute incubation in 90–100% methanol on ice. See Staining Intracellular Antigens for Flow Cytometry, Protocol C.