Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| L-Arginine hydrochloride | 211.0 | 504.0 | 2.3886256 |
| L-Cystine | 240.0 | 96.0 | 0.4 |
| L-Glutamine | 146.0 | 870.0 | 5.9589043 |
| L-Histidine hydrochloride-H2O | 210.0 | 168.0 | 0.8 |
| L-Isoleucine | 131.0 | 208.0 | 1.5877863 |
| L-Leucine | 131.0 | 208.0 | 1.5877863 |
| L-Lysine hydrochloride | 183.0 | 290.0 | 1.5846995 |
| L-Methionine | 149.0 | 60.0 | 0.40268457 |
| L-Phenylalanine | 165.0 | 128.0 | 0.77575755 |
| L-Threonine | 119.0 | 192.0 | 1.6134454 |
| L-Tryptophan | 204.0 | 40.0 | 0.19607843 |
| L-Tyrosine disodium salt dihydrate | 261.0 | 208.0 | 0.79693484 |
| L-Valine | 117.0 | 155.0 | 1.3247863 |
| Vitamins | |||
| Choline chloride | 140.0 | 4.0 | 0.028571429 |
| D-Calcium pantothenate | 477.0 | 4.0 | 0.008385744 |
| Folic Acid | 441.0 | 4.0 | 0.009070295 |
| Niacinamide | 122.0 | 4.0 | 0.032786883 |
| Pyridoxal hydrochloride | 204.0 | 4.0 | 0.019607844 |
| Riboflavin | 376.0 | 0.4 | 0.0010638298 |
| Thiamine hydrochloride | 337.0 | 4.0 | 0.011869436 |
| i-Inositol | 180.0 | 8.0 | 0.044444446 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 285.0 | 2.5675676 |
| Ferric Nitrate (Fe(NO3)3"9H2O) | 404.0 | 1.0 | 0.0024752475 |
| Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 195.0 | 1.625 |
| Potassium Chloride (KCl) | 75.0 | 800.0 | 10.666667 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 8400.0 | 100.0 |
| Sodium Chloride (NaCl) | 58.0 | 12800.0 | 220.68965 |
| Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 250.0 | 1.8115942 |
| Other Components | |||
| D-Glucose (Dextrose) | 180.0 | 9000.0 | 50.0 |