Search
* note: FBS given in mL / L.
Catalog Number(s)
31672017| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| L-Arginine hydrochloride | 210.66 | 94.8 | 0.45001423 |
| L-Cystine | 240.0 | 18.0 | 0.075 |
| L-Glutamine | 146.0 | 292.0 | 2.0 |
| L-Histidine hydrochloride-H2O | 209.63 | 31.44 | 0.14997853 |
| L-Isoleucine | 131.0 | 39.0 | 0.2977099 |
| L-Leucine | 131.0 | 39.0 | 0.2977099 |
| L-Lysine hydrochloride | 182.65 | 54.795 | 0.3 |
| L-Methionine | 149.0 | 11.0 | 0.0738255 |
| L-Phenylalanine | 165.19 | 24.765 | 0.14991827 |
| L-Threonine | 119.12 | 35.73 | 0.29994962 |
| L-Tryptophan | 204.23 | 7.65 | 0.037457768 |
| L-Tyrosine | 181.19 | 27.165 | 0.1499255 |
| L-Valine | 117.15 | 35.145 | 0.3 |
| Vitamins | |||
| Choline chloride | 140.0 | 0.75 | 0.005357143 |
| D-Calcium pantothenate | 477.0 | 0.75 | 0.001572327 |
| Folic Acid | 441.0 | 0.75 | 0.0017006802 |
| Nicotinamide | 122.0 | 0.75 | 0.006147541 |
| Pyridoxal hydrochloride | 204.0 | 0.75 | 0.0036764706 |
| Riboflavin | 376.0 | 0.075 | 1.994681E-4 |
| Thiamine hydrochloride | 337.0 | 0.75 | 0.0022255194 |
| i-Inositol | 180.0 | 1.5 | 0.008333334 |
| Inorganic Salts | |||
| Magnesium Sulfate (MgSO4-7H2O) | 246.0 | 150.0 | 0.6097561 |
| Potassium Chloride (KCl) | 75.0 | 300.0 | 4.0 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 1650.0 | 19.642857 |
| Sodium Chloride (NaCl) | 58.0 | 5100.0 | 87.93104 |
| Sodium Phosphate Dibasic Dihydrate | 156.01 | 1185.0 | 7.5956674 |
| Other Components | |||
| D-Glucose (Dextrose) | 180.0 | 750.0 | 4.1666665 |
| FBS | 250.0 | Infinity | |
| Gentimicin Sulfate | 1488.8 | 39.0 | 0.026195593 |
| Heparin | 593.45 | 250.0 | 0.42126548 |
| Phenol Red | 376.4 | 7.5 | 0.019925611 |
| Phytohemagglutinin M | 90.0 | Infinity | |