Search
These compact and powerful Raman analyzers deliver accurate, actionable results enabling user-friendly, real-time process monitoring and control.
The MarqMetrix All-In-One Process Raman Analyzer delivers reliable Raman data in real-time. You can use this solid-state analyzer across a variety of applications and with many sample types including solids, liquids, gases, powders, slurries, and heterogenous mixtures.
About the size of a textbook and with the ability to operate on its side, this small analyzer saves you valuable space; it’s easily installed anywhere from a crowded lab bench to process cabinets.
The MarqMetrix All-In-One X Process Raman Analyzer is a solid-state spectrometer engineered for real-time analysis and ease of use in locations that may be hazardous.
Built for safe, compliant operation in hazardous locations including Class I, Division 1, and ATEX-/IECEx Zone 0 environments with the same reliability, it delivers an inherently and intrinsically safe solution for areas where operators need to make fast decisions.
These versatile families of probes are designed with patented technology to simplify sampling methods and meet rigorous industry needs.
The MarqMetrix BallProbe Sampling Optic technology utilizes an exclusively sourced, high-grade spherical sapphire lens. The short focal length of the spherical optic allows for simple TouchRaman Immersion Technology sampling. Users simply touch the probe to the sample and collect reproducible spectra of liquids, solids, slurries, powders, and heterogeneous mixtures. The MarqMetrix BallProbe Sampling Optic allows non-specialists to collect high-quality data and eliminates user-to-user variation.
The Thermo Scientific MarqMetrix Process BallProbe Sampling Optic is engineered for the most demanding environments. Constructed from Hastelloy C-276, this robust probe is designed to withstand exposure to harsh chemicals and extreme conditions such as high temperature, pressure, and corrosive conditions.
This probe is available in two standard diameters and various lengths for reliable sampling across rigorous environments.
The Thermo Scientific MarqMetrix Performance BallProbe Sampling Optic is built for moderate chemical environments in both laboratory and industrial settings. Featuring a large spherical sapphire ball lens and 316L stainless steel construction, it allows for easy and reproducible Raman measurements.
Available in three standard diameters and various lengths, this probe offers versatile and reliable performance.
The Thermo Scientific MarqMetrix BioReactor BallProbe Sampling Optic is purpose-built to monitor common bioreactor processes. Durable and autoclavable past 30+ autoclave cycles, this probe includes a standard Pg 13.5 threaded adapter for universal integration into bioreactor platforms such as the Thermo Scientific HyPerforma and DynaDrive bioreactor platforms.
Built from 316L stainless steel, this probe allows for easy and reproducible Raman measurements across all sizes of bioreactors in your workflow. It’s available in multiple lengths for analysis in any bioreactor system from benchtop to production volumes.
The MarqMetrix Single-Use BioReactor BallProbe Sampling Optic is specifically designed to monitor common bioreactor processes. This probe includes a standard Pg 13.5 threaded adapter for universal integration into bioreactor platforms such as the Thermo Scientific HyPerforma and DyndaDrive bioreactor platforms.
Built from 316L stainless steel, this probe allows for easy and reproducible Raman measurements across all sizes of bioreactors in your workflow in an easy to use single-use setup. The probe is available in multiple lengths for analysis in any bioreactor system from benchtop to production volumes.
This MarqMetrix Single Use BioReactor BallProbe Sampling Optic is ideal for biopharmaceutical and pharmaceutical manufacturing due to its accurate and reproducible measurements. The probe, which comes in tear-away bags, can be sterilized using an autoclave, steam-in-place (SIP), or EtO, or it can be incorporated into larger single-use systems and gamma sterilized. The sterile probe can easily be connected to the Raman system's fiber optic interface, enabling powerful real-time chemical insights. When the process is completed, the probe can simply be disposed of to ensure batch purity.
The MarqMetrix Threaded BallProbe enables direct, in-line Raman measurement during extrusion processes. Designed for high-temperature, high-pressure environments, it threads into standard Dynisco™ ports (1/2”-20 UNF) to deliver rugged, reliable performance. Ideal for polymer, pharmaceutical, and battery material processing, it provides continuous, real-time process insight where precision and durability matter most.
The Thermo Scientific MarqMetrix FlowCell BallProbe Sampling Optic enables precise Raman measurements in a continuous flow setup. This probe is designed for seamless integration with industry-standard connections.
This probe is available in multiple flow path diameters and materials of construction, enabling integration in challenging process flow-through applications.
The Thermo Scientific MarqMetrix Proximal BallProbe Sampling Optic enables precise, real-time process monitoring without direct sample contact. This probe is ideal for samples in motion on conveyor belts and through clear physical barriers such as glass vials or windows. The specially engineered sapphire lens ensures a strong and clear Raman signal.
This probe is available in multiple sizes for different focal distances, enabling easy integration into processes without the need for contact.
For Research Use Only. Not for use in diagnostic procedures.