Search
Applied Biosystems microarray instruments provide a fully integrated platform for carrier screening research to support you with reliable and reproducible results. Our instruments combine advanced technology with user-friendly features for your microarray scanning needs to help improve throughput and efficiency in your genetic research.
Thermo Fisher Scientific offers powerful microarray solutions that are powered with robust workflows that make it easy to obtain consistent, reliable, and high-quality results critical to carrier screening research.
Our complete microarray platform includes assays, automated and manual target preparation options, fully kitted reagents, an array processing instrument, and data analysis software to enable simplified interpretation and reporting. These tools support laboratories to help maximize operational time.
The GeneTitan MC Fast Scan Instrument is a hands-free, automated solution for testing genome-wide SNP genotyping using Applied Biosystems array plates. It automates array processing from target hybridization to data generation by combining a hybridization oven, fluidics processing, and state-of-the art imaging device into a single bench-top instrument.
Key benefits are:
Thermo Fisher Scientific has partnered with Hamilton Robotics to provide you with the latest in ultra-compact and precision-based liquid-handling instrumentation. The NIMBUS Target Preparation Instrument features high-end manufacturing and exceptional pipetting performance at an affordable price.
The SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis (CE) system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis.
| GeneTitan MC Fast Scan Instrument |
|
| Dimensions | |
| 55” W x 33” D x 26” H (139.7 cm W x 83.82 cm D x 66 cm H) | |
| Application | |
| Microarray processing | |
| Weight | |
| 147 kg | |
For Research Use Only. Not for use in diagnostic procedures.