Search
Quantification can be achieved through sectioning and staining with Masson’s Trichrome, which stains the basement membrane extract blue and the endothelial cells/vessels red, or by measuring the amount of hemoglobin contained in the plug.¹,²
The primary advantage of this assay is that it is relatively easy to perform; however, it does still require the use of animals which can limit the size throughput. While the subcutaneous locale is a practical choice, it is not ideal since angiogenesis does not typically occur in these areas. The investigator should also note that current measurement techniques are not able to completely compensate for the inherent variability between animals.
Materials
LT173 updated 7-Oct-2011
For Research Use Only. Not for use in diagnostic procedures.