HCS Mitochondrial Health kit
Mitotoxicity and cytotoxicity cell health assay
The HCS Mitochondrial Health Kit simultaneously and quantitatively measures two important cell-health parameters: mitotoxicity and cytotoxicity. The MitoHealth stain detects changes in the mitochondrial membrane potential, whereupon the reagent accumulates in active mitochondria. The kit includes the Image-iT DEAD Green Stain to detect cytotoxicity, an impermeant dye to healthy cells that can enter the cells when the plasma membrane of cells is compromised.
This protocol can be used for:
  • Identifying changes in the mitochondrial membrane potential
  •  Identifying dead cells using high-content imaging
This protocol should not be used for:
  • Flow cytometry
You will need the following for this protocol:
Protocol
1. Add 30 μL DMSO to the tube of lyophilized MitoHealth stain to solubilize it (this step is needed the first time you use the kit only)
2. Culture cells in 100 µL per well of appropriate growth medium
3. Add 25 µL of test compound or drug to cells and incubate for the desired period of time under culture conditions
4. Prepare the cell staining solution by adding 10.5 μL of the MitoHealth stain (Component B) and 2.1 μL of Image-iT DEAD Green Stain (Component A) to 6 mL complete medium
5. Add 50 µL of the cell staining solution per well and incubate for 30 minutes under culture conditions—total volume should be 175 µL at this point
6. Add 3 mL 16% paraformaldehyde and 6 μL of Hoechst 33342 (Component C) to 9 mL PBS to create counterstain/fixation solution
7. Gently remove medium from each well
8. Add 100 µL of counterstain/fixation solution per well
9. Incubate for 15 minutes at room temperature
10. Gently remove counterstain/fixation solution
11. Wash wells once with 100 µL PBS
12. Add 200 µL PBS
13. Image and analyze the results
Protocol tips
  • Allow cells to grow to 80% confluence before treatment
  • Compatible with fixation, but not required
  • Dye stock solutions should be stored frozen
  • Thaw all kit components to room temperature before use
  • DMSO concentrations higher than 0.5% should not be used with live cells
Spectral information
Hoechst 33342Image-iT DEAD Green StainMitoHealth Stain
DAPI filter cube excitation
357/44 nm & emission 447/60 nm
GFP filter cube excitation
470/22 nm & emission 510/42 nm
RFP filter cube excitation
531/40 nm & emission 593/40 nm
Ordering information
Catalog # Name Size Price (USD) Qty
H10295 HCS Mitochondrial Health Kit Each
562.00