Overview
These firmware upgrades are for the CTS Xenon Electroporation System only: Gibco CTS Xenon Electroporation Instrument (Cat. No A50301).
Firmware versions will include all enhancements of previous releases in addition to the new features listed below. Updates will not in any way affect the performance of your CTS Xenon system except for the specific changes outlined below. Links and release notes for all versions are included below. We recommend updating your system to the latest version for optimized performance.
CTS Xenon System Firmware
VersionSoftware releaseRelease note
1.3.0DownloadDownload
1.2.0DownloadDownload
1.1.0DownloadDownload
1.0.6DownloadDownload
1.0.4Downloadsee below
1.0.3Downloadsee below
1.0.2Downloadsee below
1.0.1Downloadsee below
New features in Firmware 1.0.4
  1. Update firmware for precool subsystem
  2. Update OPC UA server with new information
New features in Firmware 1.0.3
  1. Added PDF report generation
  2. Added sample retrieval workflow
  3. Prevent user from starting extraction if protocol selected is not created by signed in user
  4. Prevent user sign out in CM mode during a run. The sign out will only happen when the run is completed
  5. Added new information for OPC server
New features in Firmware 1.0.2
  1. Added Open Platform Communication features
  2. Fixed manual purge issues
New features in Firmware 1.0.1
  1. Added skip extraction
  2. Added manual drain purge
  3. Added FSE Test Suite methods
  4. Fixed WIFI WPA and WEP hexadecimal issues
Downloading the firmware file
  1. Download the attached file onto a USB memory stick (shown in .update format).
    Important: For Internet Explorer users, right click the above “download” link. Click “Save Target As”. Change the file extension to “.update” and click save;
  2. Save/Transfer the .update file (not in any folder) on your USB stick.
  3. Make sure that no other .update files are present on the memory stick.
Updating your CTS Xenon instrument
  1. Power on the instrument.
  2. Insert the USB memory stick containing the firmware update into one of the USB ports in the CTS Xenon instrument.
  3. On the Home screen, touch ‘Settings’.
  4. In the Settings menu, touch ‘Maintenance & Services’.
  5. Touch 'Software Updates’.
  6. If you have downloaded the firmware properly, the file will appear on the screen.
  7. Select the file on the screen, and press 'Upgrade' at the bottom of the screen. 
  8. Touch ‘Yes’ to confirm that you want to upgrade.
    IMPORTANT! Ensure that you do not turn off the instrument when the upgrade is in progress.
  9. When the upgrade process is complete, the instrument will restart automatically. When it has rebooted, you can remove your USB stick.
LEGAL STATEMENTS
Important licensing information:
DISCLAIMER: TO THE EXTENT ALLOWED BY LAW, THERMO FISHER SCIENTIFIC INC. AND/OR ITS AFFILIATE(S) WILL NOT BE LIABLE FOR SPECIAL, INCIDENTAL, INDIRECT, PUNITIVE, MULTIPLE, OR CONSEQUENTIAL DAMAGES IN CONNECTION WITH OR ARISING FROM THIS DOCUMENT, INCLUDING YOUR USE OF IT.