CTS™ Xenon™ Electroporation System
Gibco™

CTS™ Xenon™ Electroporation System

The Gibco CTS Xenon Electroporation System enables reliable delivery of DNA, RNA, proteins, and other payloads into cells, with exceptionalRead more
Have Questions?
Catalog NumberQuantity
A503011 system
Catalog number A50301
Price (USD)
-
Quantity:
1 system

The Gibco CTS Xenon Electroporation System enables reliable delivery of DNA, RNA, proteins, and other payloads into cells, with exceptional cell viability and recovery. This leads to efficient transfection and genetic modification, even in hard-to-transfect cell types.

Features of the CTS Xenon Electroporation System include:
• High speed and large volume—transfect up to 2.5 x 109 T cells/25 mL in less than 25 minutes
• Scalable, proven performance and viability—up to 90% gene knockout and 80% cell viability
• Process flexibility—user-programmable system enables you to create and optimize electroporation protocols for various cell types and payloads, from process development through commercial manufacturing
• Efficient nonviral transfection—deliver DNA, RNA, and protein payloads, including CTS TrueCut Cas9 Protein
• Closed-system processing—Xenon MultiShot Electroporation Cartridge enables sterile welding to PVC or C-Flex™ tubing

A choice of two single-use electroporation chambers of different volume capacities provides flexibility: the 1 mL SingleShot chamber enables process development while the 5-25 mL MultiShot cartridge can be used in a closed, commercial-scale manufacturing system that minimizes contamination and human error while maximizing safety.

In conjunction with the Neon Transfection System, the CTS Xenon Electroporation System helps enable smooth scale-up from research to process development to commercial manufacturing of up to 25 mL of cells at a time. Paired with other Thermo Fisher Scientific instruments, the Xenon system can be used to assemble a closed, integrated, GMP-compliant workflow that can be controlled by process automation software.

Learn more about the CTS Xenon Electroporation System
View the interactive 3D product tour
Request a demo

Specifications
Dimensions (L x W x H)27.4 x 26.5 x 21.2 in. (Door open), 43.1 x 26.5 x 21.2 in. (Door closed)
Electrical Requirements100/240 VAC
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
Product TypeElectroporator
Pulse Number Range1 to 10
Pulse Width Range1 to 30 ms
Quantity1 system
Voltage Range (HV Mode)500 to 2500 V
Volume (Metric)1 mL, 5–25 mL (Input)
Weight (English)154 lb.
Weight (Metric)70 kg
DisplayGraphical User Interface
InterfaceTouchscreen
Unit SizeEach