One Lambda One Lambda™ | Product Documentation & Support
Order Status Order Status Login | Register | Log Out
Search
clear search
Recent searches Clear History
Suggestions
All Results
Search
HLA Typing
Micro SSP™ Typing Trays
PCR-SSP designed to broaden typing coverage and reduce ambiguities
Broaden Typing Coverage and Reduce Ambiguities
One Lambda’s portfolio of Micro SSP typing trays provide complete HLA Class I and Class II DNA typing. Using Generic Trays, technologists can either assign HLA Class I or Class II alleles individually or simultaneously. If more detailed typing is needed, the High-Resolution Allele Specific Trays will help eliminate redundancies.
Micro SSP Generic Trays
One Lambda's Generic trays are available to assign either HLA Class I or Class II alleles, or Class I and Class II alleles simultaneously. These basic molecular trays use a 96-well format (unless otherwise stated) with pre-aliquoted and dispensed primers and an internal control in each well.
  • Pre-aliquoted and dispensed primers in a 96-well format save time and are easy to use 
  • Optimized reagents are ready to use
  • Internal control included in each well
  • Comprehensive allele coverage
  • Easy gel interpretation
  • Fast results in less than two hours
Micro SSP Allele Specific Trays
Our Micro SSP Allele Specific HLA Class I and Class II Trays are designed for fast and accurate HLA Class I and Class II sub-typing. These trays can be used alone or in combination with One Lambda’s Micro SSP Generic and High Resolution HLA Typing Trays.
  • Pre-aliquoted and dispensed primers in a 96-well format save time and are easy to use
  • Optimized reagents are ready to use
  • Internal control included in each well
  • Comprehensive allele coverage
  • Easy gel interpretation
  • Fast results in less than two hours
Micro SSP High Resolution Trays
The Micro SSP High Resolution Trays offer more detailed subtyping information for Class II alleles (DPA1/DPB1, DQA1 and DRB1,3, 4, 5). They allow for focused testing eliminating ambiguities, and confirm questionable results.
  • Pre-aliquoted and dispensed primers save time and are easy to use
  • Specific—provides detailed allelic typing
  • Economical—allows for focused testing, eliminating test redundancy
  • Allows confirmation of questionable or ambiguous typings
  • Fast results in less than two hours
SSP B27 Primer Sets
These primer sets use SSP technology and allow customers to configure their own typing tests and to use only the primers they need to type for B27 alleles.
  • Uses the same familiar technology as One Lambda’s Micro SSP products.
  • B27 primers - can be configured to use only the primers needed
It all starts with a demo
Click here to request a demo for Micro SSP™
Related Products
HLA Fusion™ Software
Analysis results for Antibody Detection and Molecular Typing products.
96 Well Transfer Device
The 96-Well Transfer Device uses 96 pipettes to transfer reagents quickly and simultaneously across the tray, ensuring consistency throughout the array