Search
View additional product information for eBioscience™ TMB Solution (1X) - FAQs (00-4201-56)
3 product FAQs found
No, we offer different TMB substrates for different applications.
We recommend the following TMB substrates for this particular application:
- eBioscience TMB Solution (1X) (Cat. No. 00-4201-56)
- (For higher sensitivity) 1-Step Ultra TMB-ELISA Substrate Solution (Cat. No. 34029)
TMB is a faster developing, stronger substrate that yields greater amplification/sensitivity/background. The use of an acid stop solution produces a yellow end product read at 450nm. Super AquaBlue is a slower developing blue/green substrate particularly useful for kinetic studies.
When working with our matched pairs or uncoated ELISA kits, please refer to the user guide provided with your antibody-matched pair for kit-specific guidance. When designing your own ELISA, please utilize our ELISA Builder tool for reagent selection as well as our ELISA Development and Optimization pages for futher guidance.