Zinc ingot, 99.999% (metals basis) - FAQs
View additional product information for Zinc ingot, 99.999% (metals basis) - FAQs (000420.KF)
1 product FAQs found
How much does a single piece (1 pc.) of Zinc ingot 99.999% (metals basis) weigh?
A single piece (1 pc.) of Zinc ingot 99.999% (metals basis) is approximately equal to 200 g.