Zinc ingot, 99.999% (metals basis)
Zinc ingot, 99.999% (metals basis)
Zinc ingot, 99.999% (metals basis)
Thermo Scientific Chemicals

Zinc ingot, 99.999% (metals basis)

CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Catalog NumberQuantity
000420.KF
also known as 000420-KF
1 Pc.
Catalog number 000420.KF
also known as 000420-KF
Price (USD)
972.65
Online Exclusive
1,081.00
Save 108.35 (10%)
Each
Quantity:
1 Pc.
Request bulk or custom format
Price (USD)
972.65
Online Exclusive
1,081.00
Save 108.35 (10%)
Each
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Appearance (Color)Gray
FormIngot
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

How much does a single piece (1 pc.) of Zinc ingot 99.999% (metals basis) weigh?

A single piece (1 pc.) of Zinc ingot 99.999% (metals basis) is approximately equal to 200 g.