Anti-Clumping Agent, 100 mL - FAQs
View additional product information for Anti-Clumping Agent - FAQs (0010057DG, 0010057AE)
1 product FAQs found
What reagents do you offer for cell dissociation, and what are the main differences between them?
Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).
Find additional tips, troubleshooting help, and resources within ourMammalian Cell Culture Basics Support Center.