Product Image
Applied Biosystems™
GeneTitan™ MC Xenon Lamp
GeneTitan™ MC Xenon Lamp is an instrument accessory for GeneTitan™ MC Instrument North America/Japan (110V) (Cat no. 00-0372) and GeneTitan™ MC Instrument International (220V) (Cat no. 00-0373).
Have Questions?
Catalog NumberQuantity
01-07401 Ea.
Catalog number 01-0740
Price (MXN)
-
Quantity:
1 Ea.
GeneTitan™ MC Xenon Lamp is an instrument accessory for GeneTitan™ MC Instrument North America/Japan (110V) (Cat no. 00-0372) and GeneTitan™ MC Instrument International (220V) (Cat no. 00-0373). The imaging device in the GeneTitan MC Instrument uses an external, high-intensity xenon lamp and dual excitation and emission filters to capture images from array plates for use in expression, genotyping, and microbiome studies.
Related Links:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)GeneTitan Multi-Channel (MC) Instrument
Quantity1 Ea.
For Use With (Application)GeneTitan Multi-Channel (MC) Instrument Accessory
Unit SizeEach
Frequently asked questions (FAQs)
What is the catalog number for the GeneTitan MC Xenon Lamp?
The catalog number for the GeneTitan MC Xenon Lamp is 01-0740.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.