Search
View additional product information for 123count eBeads™ Counting Beads - FAQs (01-1234-42)
3 product FAQs found
The first thing to do is check your threshold and see if it is set on forward scatter. If so, the beads are probably being excluded by the threshold. Reducing the threshold setting should reveal your beads.
Cell counting using flow cytometry can be accomplished by adding an internal microsphere counting standard to the flow cytometric sample. The number of reference beads that are collected reflects a known volume. This allows you to calculate cell concentration.
Yield is the percentage of target cells harvested based on the number of cells theoretically available in the starting population. To calculate yield, stain a sample of unsorted and sorted cells with antibodies to identify the target cells, and count the total number of target cells using 123count eBeads (Cat. No. 01-1234-42) on the cytometer. Yield = # of sorted target cells, # of target cells unsorted.
Purity is the percentage of cells harvested that are target cells. Purity can be directly generated by flow cytometric analysis of sorted samples. To obtain purity, stain the sorted cells with antibodies to identify the target cells and analyze by flow cytometry. Gate on live, single cells, and subsequently gate on the target cell population. Purity is the percentage of target cells in the live, single-cell gate.