Fiberlite 250 mL Bottle System, PPCO (set of 6) - FAQs
Fiberlite 250 mL Bottle System, PPCO (set of 6) - FAQs
View additional product information for Fiberlite 250 mL Bottle - FAQs (010-1496, 010-1495)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.