SteriBIOS 1000 mL Bottle, PPCO - FAQs
View additional product information for SteriBIOS 1000 mL Bottle, PPCO - FAQs (010-1661)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.