Search
View additional product information for Sodium ingot, 99.8% (metals basis) - FAQs (010342.60, 010342.35)
1 product FAQs found
Sodium ingot, 99.8% (metals basis) is packaged dry (not in oil) in a plastic bag, flushed with argon, and placed in a metal can.