Frequently asked questions (FAQs)
How is Sodium ingot, 99.8% (metals basis) packaged?
Sodium ingot, 99.8% (metals basis) is packaged dry (not in oil) in a plastic bag, flushed with argon, and placed in a metal can.