Bromure de potassium, ACS, 99 % min., Thermo Scientific Chemicals
Bromure de potassium, ACS, 99 % min., Thermo Scientific Chemicals
Bromure de potassium, ACS, 99 % min., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bromure de potassium, ACS, 99 % min., Thermo Scientific Chemicals

Pour la fabrication de papiers photographiques, la photogravure en couleur et la lithographie, la spectroscopie
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
50 g
250 g
1 kg
5 kg
Référence 012612.18
également connu sous le numéro 012612-18
Prix (EUR)
28,70
Each
Quantité:
50 g
Grand volume ou format personnalisé
Prix (EUR)
28,70
Each
Identifiants chimiques
CAS7758-02-3
IUPAC Namepotassium bromide
Molecular FormulaBrK
InChI KeyIOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES[K+].[Br-]
Voir plus
Appearance (Color)White
FormCrystals or crystalline powder or solid chunks
Assay (Argentometric Titration)≥99.0%
Barium content≤0.002%
Bromate content≤0.001%
Voir plus
Used in manufacturing of photographic papers, engraving and lithography, in spectroscopy, and as an analytical lab reagent. Potassium bromide is applied as a veterinary drug, as an antiepileptic medication for dogs. It is utilized for the bromination of alkenes using cerium(IV) ammonium nitrate. It plays an essential role with diacetoxy iodobenzene for the efficient bromination of alkenes and alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only