Sulfate d’hydrogène de potassium, fusionné, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfate d’hydrogène de potassium, fusionné, ACS, Thermo Scientific Chemicals

Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
100 g
500 g
Référence 013800.22
également connu sous le numéro 013800-22
Prix (EUR)
66,50
Each
Quantité:
100 g
Grand volume ou format personnalisé
Prix (EUR)
66,50
Each
Spécifications
AspectWhite fused powder
Nom chimique ou matériauPotassium hydrogen sulfate, fused
CouleurBlanc
Informations DOTClasse de risque :8, Groupe d’emballage :II
Poids de la formule136.17
Voir plus
Potassium hydrogen sulfate is used in the conversion of tartrates to bitartrates in wine. It is also used as a disintegrating agent in analytical chemistry. Further, it is used to make fertilizers and in the preparation of potassium persulfate. In addition to this, it is used as a catalyst in organic preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only