Phosphate de magnésium pentahydraté, P<sub>2</sub>O<sub>5</sub> 33 %, Thermo Scientific Chemicals
Phosphate de magnésium pentahydraté, P<sub>2</sub>O<sub>5</sub> 33 %, Thermo Scientific Chemicals
Phosphate de magnésium pentahydraté, P<sub>2</sub>O<sub>5</sub> 33 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phosphate de magnésium pentahydraté, P2O5 33 %, Thermo Scientific Chemicals

CAS: 10233-87-1 | H10Mg3O13P2 | 352.93 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
1 kg
5 kg
Référence 018219.A7
également connu sous le numéro 018219-A7
Prix (EUR)
216,00
Each
Quantité:
5 kg
Grand volume ou format personnalisé
Prix (EUR)
216,00
Each
Identifiants chimiques
CAS10233-87-1
IUPAC Namemagnesium(2+) pentahydrate phosphate
Molecular FormulaH10MgO9P
InChI KeyZHDVAZVXDZNFPS-UHFFFAOYSA-K
SMILESO.O.O.O.O.[Mg++].[O-]P([O-])([O-])=O
Voir plus
Assay (unspecified)P_2O_5 33% min
Magnesium phosphate pentahydrate is used in the preparation of magnesium calcium phosphate biocement(MCPB) for bone regeneration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only