Thermo Scientific Chemicals

1-chlorure de n-butylpyridinium, 98 %, Thermo Scientific Chemicals

Référence: 019740.09
10 g, Each
Thermo Scientific Chemicals

1-chlorure de n-butylpyridinium, 98 %, Thermo Scientific Chemicals

Référence: 019740.09
10 g, Each
Quantité
Référence: 019740.09
également connu sous le numéro 019740-09
Prix (EUR)
Quantité
-

Identifiants chimiques

CAS
1124-64-7
IUPAC Name
1-butylpyridin-1-ium
Molecular Formula
C9H14N
InChI Key
REACWASHYHDPSQ-UHFFFAOYSA-N
SMILES
CCCC[N+]1=CC=CC=C1
Solution Test
In water: Clear
Melting Point (clear melt)
130.0-136.0?C
Assay (Titration ex Ag)
≥97.5 to ≤102.5%

Description

The derivatives of 1-n-Butylpyridinium chloride is used in spent fuel reprocessing, particularly in the extraction of high-level radioactive aqueous waste from the processing of nuclear fuel. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text