Search
Buscar
View additional product information for Al-24 Tube (one end closed);OD (mm), 24;ID (mm), 18 - FAQs (030093.DR, 030093.DW)
1 product FAQs found
The maximum working temperature for the Al-24 Tube is 1950 degrees C.