Al-24 Tube (one end closed);OD (mm), 24;ID (mm), 18
Thermo Scientific Chemicals
Al-24 Tube (one end closed);OD (mm), 24;ID (mm), 18
Have Questions?
Change viewbuttonViewtableView
Quantity:
200 mm
300 mm
Catalog number 030093.DR
also known as 030093-DR
Price (USD)
196.65
Online Exclusive
218.00
Save 21.35 (10%)
Each
Quantity:
200 mm
Request bulk or custom format
Price (USD)
196.65
Online Exclusive
218.00
Save 21.35 (10%)
Each
Specifications
Chemical Name or MaterialAl-24 Tube
Name NoteOne end closed
Recommended StorageAmbient temperatures
TSCAU
Quantity200 mm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Frequently asked questions (FAQs)
What is the maximum working temperature of Al-24 Tubes?
The maximum working temperature for the Al-24 Tube is 1950 degrees C.