Al-23 Solid Rod, Round;Diameter (mm), 6 - FAQs
View additional product information for Al-23 Solid Rod, Round, Dia.: 6mm, Thermo Scientific Chemicals - FAQs (032933.DR)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.