Al-23 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 75 x 50 x 44;Vol (ml), 110 - FAQs
Al-23 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 75 x 50 x 44;Vol (ml), 110 - FAQs
View additional product information for Al-23 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 40 x 30 x 26;Vol (ml), 20 - FAQs (032952.KF, 032951.KF, 032953.KF, 032955.KF, 032954.KF)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.