Al-24 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 100 x 65 x 55;Vol (ml), 225 - FAQs
Al-24 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 100 x 65 x 55;Vol (ml), 225 - FAQs
View additional product information for Al-24 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 40 x 30 x 26;Vol (ml), 20 - FAQs (032966.KF, 032961.KF, 032964.KF, 032965.KF, 032962.KF, 032963.KF)
1 product FAQs found
What is the maximum working temperature of Al-24 Tubes?
The maximum working temperature for the Al-24 Tube is 1950 degrees C.