Aluminon, ACS - FAQs
Aluminon, ACS - FAQs
View additional product information for Aluminon, ACS - FAQs (036239.14, 036239.22)
1 product FAQs found
What is the solubility of Aluminon?
Aluminon is soluble in water at a concentration of ~800 g/L at 20°C. It is slightly soluble in ethanol and insoluble in petroleum ether.