Sel disodique d’acide éthylènediaminetetraacétique, solution Std. 0,200N (0,1M), Thermo Scientific Chemicals
Sel disodique d’acide éthylènediaminetetraacétique, solution Std. 0,200N (0,1M), Thermo Scientific Chemicals
Sel disodique d’acide éthylènediaminetetraacétique, solution Std. 0,200N (0,1M), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sel disodique d’acide éthylènediaminetetraacétique, solution Std. 0,200N (0,1M), Thermo Scientific Chemicals

CAS: 139-33-3 | C10H14N2Na2O8 | 336.21 g/mol
Have Questions?
RéférenceQuantité
039783.K2
également connu sous le numéro 039783-K2
1 L
Référence 039783.K2
également connu sous le numéro 039783-K2
Prix (EUR)
54,30
Each
Quantité:
1 L
Grand volume ou format personnalisé
Prix (EUR)
54,30
Each
Identifiants chimiques
CAS139-33-3
IUPAC Namedisodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
Molecular FormulaC10H14N2Na2O8
InChI KeyZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Voir plus
Appearance (Color)Clear colorless
FormLiquid
Concentration0.200N ± 0.19% (0.0019N)
Ethylenediaminetetraacetic acid disodium salt is widely used in textile industry. Usually applied to dissolve limescale. It is applied in textile industry, pulp and paper industry and also in chelation therapy. In cosmetics, it acts as a sequestering agent. It acts as a corrosion inhibitor to carbon steel in the industries. It also acts as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only