Feuille d’acier inoxydable, 0,5 mm (0,02 po) d’épaisseur, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Feuille d’acier inoxydable, 0,5 mm (0,02 po) d’épaisseur, Thermo Scientific Chemicals

Fe:Cr:Ni; 70:19:11 wt%
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Référence 041585.FI
également connu sous le numéro 041585-FI
Prix (EUR)
51,20
Each
Quantité:
50 x 50 mm
Grand volume ou format personnalisé
Prix (EUR)
51,20
Each
Spécifications
AspectSilver -colored foil
Nom chimique ou matériauStainless Steel
CouleurArgent
Numéro MDLMFCD00148499
Formule moléculaireFe:Cr:Ni; 70:19:11 wt%
Voir plus
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only