N-Hexane, qualité environnementale, >95 %, Thermo Scientific Chemicals
N-Hexane, qualité environnementale, >95 %, Thermo Scientific Chemicals
N-Hexane, qualité environnementale, >95 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Hexane, qualité environnementale, >95 %, Thermo Scientific Chemicals

Pour l’analyse des résidus de pesticides, de l’environnement et du GC
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
1 L
4 x 1 L
Référence 042100.K2
également connu sous le numéro 042100-K2
Prix (EUR)
55,20
Each
Quantité:
1 L
Grand volume ou format personnalisé
Prix (EUR)
55,20
Each
Identifiants chimiques
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
Voir plus
Impurity content≤10ppt Heptachloepoxide ( as single peak GC-ECD responsive residue)
Assay (GC)≥95.0% ( n-Hexane)
Residue on Evaporation≤2ppm
Color (APHA)≤10
Assay (GC)≥99.5% (as C6 Hydrocarbons)
Voir plus
Pesticide residue, environmental and GC analysis.n-Hexane is used in spectrophotometry and environmental testing. It is used as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. other uses are as Fuels and fuel additives, Intermediates and Laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only