Thermo Scientific Chemicals

Sodium chlorate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 044408.36
500 g, Each
Thermo Scientific Chemicals

Sodium chlorate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 044408.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 044408.36
also known as 044408-36
Price (USD)
Price: 95.00
Your price:
Quantity
-

Chemical Identifiers

CAS
7775-09-9
IUPAC Name
sodium chlorate
Molecular Formula
ClNaO3
InChI Key
YZHUMGUJCQRKBT-UHFFFAOYSA-M
SMILES
[Na+].[O-][Cl](=O)=O
Assay (unspecified)
99.0% NaClO₃ min
Impurity content
BrO₃ 0.015%
Impurity content
Ca 0.002%
Impurity content
Cl 0.005%
Impurity content
Fe 5ppm

Description

Sodium chlorate is used to prepare chlorine dioxide in the pulp and paper industry. It is utilized for bleaching the wood pulp in order to make environmentally friendly and quality white paper products. It is also used to produce other perchlorate compounds in the industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text